Compare Ion Chromatography Systems
Which ion chromatography system is right for my laboratory?
It depends. Are you running routine analyses or do you need advanced functions? You can choose from basic workhorses to high-pressure systems with electrolytic eluent generation. What’s the difference and how do they compare? Use the table below to see which one is right for you.
Speak to a specialist
  Dionex Aquion IC System Dionex Integrion HPIC System Dionex ICS-4000 Capillary HPIC System Dionex ICS-6000 HPIC System
  Dionex Aquion IC System Dionex Integrion HPIC System Dionex ICS-4000 Capillary HPIC System Dionex ICS-6000 HPIC System
  Request quote Request quote Request quote Request quote
Advantages Simple analyses Routine analyses Always ready, routine analyses Routine and complex analyses
Automated, comprehensive consumables tracking reduces installation & tracking errors    
Reusable, ultralow-dead-volume, high-pressure finger-tight connections that are easy to use      
Accurate, reproducible electrolytic high-pressure gradients with shorter run times      
Easily track suppressor performance using single-device suppression        
Single-vendor software that controls multiple vendor instruments        
Built-in method development software tool to efficiently optimize column and separation conditions        
Precise, accurate electrolytic eluent generation for excellent method reproducibility      
Integrated detector options that protect your investment      
Capillary IC system with the advantages of always-ready operation    
Multiple pump configurations for your application flexibility  
Runs 2D-IC applications for trace analyses  
Mechanical and RFIC gradients to expand your capabilities  
Increase productivity using single and dual configurations  
Broad flow-rate options for capillary, microbore, and standard bore columns  
Easy methods development using dual column temperature zones  
Reconfigure with individual, stand-alone modules for special application requirements  
CMD SchemaApp code