Simplicity to everyday analyses
The Thermo Scientific Dionex Easion gives you everything you need for day to day IC analyses in one small package. No additional pumps or accessories are required to operate Dionex Easion. Anion and cation chemistry kits and Thermo Scientific Chromeleon CDS eWorkflows take the guess work out of set-up and operation.
Advanced suppressor technology
Worry-free suppressors increase analyte signal while decreasing background noise by removing conductive ions from the eluent. Dionex Easion is compatible with our chemical suppressors.
Our membrane-based chemical suppressors ensure maximum uptime and reduces baseline drift to minimize sample to sample variability, improving sample to sample reproducibility. Continuously regenerated, it is always ready for the next sample. No need to wash and regenerate between runs.
Flow path materials
Inert, nonmetallic PEEK components throughout the system ensure compatibility with corrosive eluents and metal-contamination-free ion chromatography. The dual-piston PEEK pump is chemically inert—engineered to handle IC eluents and reduce maintenance costs.
Simple operation and maintenance
With the Dionex Easion system, there are no extra items to change or maintain. The simple design provides outstanding access to chromatography components.
- Eluent and regenerant tray holds bottles securely to prevent spills
- Dual-piston PEEK isocratic pump
- Fixed loop injection valve accommodates either manual injection or an autosampler and can be easily changed to accommodate different injection volumes
- Worry-free chemical suppressor is continuously regenerated
- Thermally controlled conductivity detector reduces noise and improves peak area consistency
Thermo Scientific Chromeleon 7 Chromatography Data System (CDS) has the best-in-class performance and usability, unlocking the full potential of your Thermo Scientific IC system.
Chromeleon CDS was designed to be easy to use and resemble familiar office software products, to ensure minimal training time to get started. Client-server architecture and industry-leading network failure protection ensure that your operation is never interrupted.
To further reduce complexity for operators, eWorkflows are available for all our essential applications. An eWorkflow procedure is a set of rules that captures all of the unique aspects of an ion chromatography workflow, such as instrument conditions, and injection sequence requirements, and guides the operator through a minimal number of choices needed to create a complete, correct sequence with predefined files and a well-defined structure.
Autosampler-compatible
Autosamplers enable automated sample injection, and the Dionex Easion is compatible with both our Thermo Scientific Dionex AS-DV and Thermo Scientific Dionex AS-AP models. The Dionex AS-DV is recommended for walk-away convenience and built-in sample filtration.
Services for ion chromatography systems
The Unity Lab Services team is ready to support your ion chromatography (IC) instrument from installation and warranty to service plans and compliance services. Our full range of services helps ensure you get the best performance from your system throughout its life. We can help you choose the right plan and add services to best fit your needs, so your instrument operates reliably and efficiently when you need it.
World class expertise
As the world leader in serving science and part of the Thermo Fisher Scientific organization, we are ready to serve you with more than 2,000 highly trained Unity Lab Services technical professionals globally. In an organization that has been serving our customers for more than 50 years, our engineers have on average 18 years of experience operating and maintaining instruments we service.
Each field engineer who works on your instrument is OEM factory trained and certified on that specific instrument, so you can be confident in your audit-readiness with regular service. You are guaranteed to receive consistent, high-quality installation, maintenance and repair of your IC system with world-class expertise on the Unity Lab Services team.
Service plans
Your specific needs and expectations as you use your ion chromatography instrument require a level of service tailored to those needed. Choose an extended warranty when you purchase your instrument or browse our service plans to find the perfect fit.
Choosing an extended warranty gets you up to 50% faster response time and more than 30% reduced downtime. The extended warranty includes enhanced technical support enabling remote resolution of 35% of issues, priority on-site response time with a 3-business-day target response for corrective maintenance and repairs, including factory-certified parts, labor, and travel; pre-scheduled on-site preventive maintenance (PM) and PM parts kit; and discounts on training courses, parts, consumables and accessories.
Features and benefits of our most popular service plan
Features | Billable Service | Extended Warranty | Benefits |
---|---|---|---|
Annual preventive maintenance PM kit | ![]() |
![]() |
Boosted uptime |
Digital remote support | ![]() |
![]() |
More than 36% remote resolution reduces downtime Faster diagnosis and improved first-time fix |
Enhanced technical support | ![]() |
![]() |
Priority, direct access to highly experienced technical staff |
Priority on-site response time target | No target | 3 business days | Target timeline to plan for corrective maintenance and get up and running |
Corrective maintenance including parts, labor and travel | ![]() |
![]() |
Reduced repair cost |
10% discounts on parts, consumables and accessories | ![]() |
![]() |
Reduced cost for products you regularly need for your instrument |
System manuals and specifications
Advanced suppressor technology
Worry-free suppressors increase analyte signal while decreasing background noise by removing conductive ions from the eluent. Dionex Easion is compatible with our chemical suppressors.
Our membrane-based chemical suppressors ensure maximum uptime and reduces baseline drift to minimize sample to sample variability, improving sample to sample reproducibility. Continuously regenerated, it is always ready for the next sample. No need to wash and regenerate between runs.
Flow path materials
Inert, nonmetallic PEEK components throughout the system ensure compatibility with corrosive eluents and metal-contamination-free ion chromatography. The dual-piston PEEK pump is chemically inert—engineered to handle IC eluents and reduce maintenance costs.
Simple operation and maintenance
With the Dionex Easion system, there are no extra items to change or maintain. The simple design provides outstanding access to chromatography components.
- Eluent and regenerant tray holds bottles securely to prevent spills
- Dual-piston PEEK isocratic pump
- Fixed loop injection valve accommodates either manual injection or an autosampler and can be easily changed to accommodate different injection volumes
- Worry-free chemical suppressor is continuously regenerated
- Thermally controlled conductivity detector reduces noise and improves peak area consistency
Thermo Scientific Chromeleon 7 Chromatography Data System (CDS) has the best-in-class performance and usability, unlocking the full potential of your Thermo Scientific IC system.
Chromeleon CDS was designed to be easy to use and resemble familiar office software products, to ensure minimal training time to get started. Client-server architecture and industry-leading network failure protection ensure that your operation is never interrupted.
To further reduce complexity for operators, eWorkflows are available for all our essential applications. An eWorkflow procedure is a set of rules that captures all of the unique aspects of an ion chromatography workflow, such as instrument conditions, and injection sequence requirements, and guides the operator through a minimal number of choices needed to create a complete, correct sequence with predefined files and a well-defined structure.
Autosampler-compatible
Autosamplers enable automated sample injection, and the Dionex Easion is compatible with both our Thermo Scientific Dionex AS-DV and Thermo Scientific Dionex AS-AP models. The Dionex AS-DV is recommended for walk-away convenience and built-in sample filtration.
Services for ion chromatography systems
The Unity Lab Services team is ready to support your ion chromatography (IC) instrument from installation and warranty to service plans and compliance services. Our full range of services helps ensure you get the best performance from your system throughout its life. We can help you choose the right plan and add services to best fit your needs, so your instrument operates reliably and efficiently when you need it.
World class expertise
As the world leader in serving science and part of the Thermo Fisher Scientific organization, we are ready to serve you with more than 2,000 highly trained Unity Lab Services technical professionals globally. In an organization that has been serving our customers for more than 50 years, our engineers have on average 18 years of experience operating and maintaining instruments we service.
Each field engineer who works on your instrument is OEM factory trained and certified on that specific instrument, so you can be confident in your audit-readiness with regular service. You are guaranteed to receive consistent, high-quality installation, maintenance and repair of your IC system with world-class expertise on the Unity Lab Services team.
Service plans
Your specific needs and expectations as you use your ion chromatography instrument require a level of service tailored to those needed. Choose an extended warranty when you purchase your instrument or browse our service plans to find the perfect fit.
Choosing an extended warranty gets you up to 50% faster response time and more than 30% reduced downtime. The extended warranty includes enhanced technical support enabling remote resolution of 35% of issues, priority on-site response time with a 3-business-day target response for corrective maintenance and repairs, including factory-certified parts, labor, and travel; pre-scheduled on-site preventive maintenance (PM) and PM parts kit; and discounts on training courses, parts, consumables and accessories.
Features and benefits of our most popular service plan
Features | Billable Service | Extended Warranty | Benefits |
---|---|---|---|
Annual preventive maintenance PM kit | ![]() |
![]() |
Boosted uptime |
Digital remote support | ![]() |
![]() |
More than 36% remote resolution reduces downtime Faster diagnosis and improved first-time fix |
Enhanced technical support | ![]() |
![]() |
Priority, direct access to highly experienced technical staff |
Priority on-site response time target | No target | 3 business days | Target timeline to plan for corrective maintenance and get up and running |
Corrective maintenance including parts, labor and travel | ![]() |
![]() |
Reduced repair cost |
10% discounts on parts, consumables and accessories | ![]() |
![]() |
Reduced cost for products you regularly need for your instrument |
System manuals and specifications
Systems & Kits
Anion and cation chemistry kits take the guess work out of setup. All columns and consumables for your application are necessary at the time of installation. Please ensure that you order a kit or the necessary products with your instrument.
Resources
Systems
Software
Autosampler
Support
Service
We provide a full range of service options for all Thermo Scientific Ion Chromatography systems.
Featured service plans
Premier services provide our highest priority, fastest response to maximize uptime.
Extended Warranty gives you 50% faster response time and 30% reduced downtime compared to billable.
Additional services
Annual Preventative Maintenance is a core component of our service plans, but we also offer it as a standalone service.