Stabilize retention time with precise thermostatting
Stabilize retention time with precise thermostatting
Whether you have simple or complex method requirements, Thermo Scientific™ Vanquish™ and UltiMate™ 3000 HPLC/UHPLC system column compartments ensure accurate and precise temperature control for your columns, as well as for optional valves for 2D-LC or application switching.
LC column compartments
Column Compartment
Feature
Vanquish™ Column Compartment H Control separations with more confidence using an entirely new and unique combination of active or passive eluent preheating and advanced column thermostatting.
UltiMate™ 3000 TCC-3000SD Standard Thermostatted Column Compartments Ensure accurate and precise temperature control from 5° to 80°C for your standard flow rate applications. Optional valves enable 2D-LC or application switching.
UltiMate™ 3000 TCC-3000RS Rapid Separation Thermostatted Column Compartment Optimize column selection for method development and ensure accurate and precise temperature control for your existing methods all the way to 110°C.
UltiMate™ 3000 ACC-3000 Autosampler Column Compartments Save money and space with this autosampler with integrated column compartment which supports conventional HPLC and UHPLC at backpressures up to 62MPa.
Featured column temperature control video
Using Vanquish to Control Column Temperature
Evert-Jan Sneekes, HPLC Strategic Marketing Manager, helps us understand how we can use Vanquish to control column temperature.
Are you using the right LC column?
Access the latest version of our Chromatography Columns and Consumables Catalog in PDF and online formats. The enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog ›