Sensitive, universal detection
Sensitive, universal detection
Obtain sensitive, universal detection with a near-uniform response. Charged aerosol detectors measure a wide range of analytes in the areas of pharmaceuticals (large and small molecule), biomolecules, foods and beverages, specialty chemicals, and polymers. Their flexibility and performance are ideal for analytical R&D, while the detectors' simplicity and reproducibility benefit manufacturing QC/QA applications.
HPLC and UHPLC charged aerosol detectors
Corona Veo Charged Aerosol Detectors
For use with capillary, microbore and normal-bore columns to take advantage of ever advancing column technologies. The Corona Veo detector is designed to integrate into any LC, HPLC or UHPLC system from any manufacturer.
Utilize the increased separation speed and resolution of the Vanquish UHPLC system to detect and quantify virtually any substance using the near-universal detection capabilities of Vanquish Charged Aerosol Detectors.
Vanquish Charged Aerosol Detectors
Featured videos
Charge aerosol detection bibliography
See how other labs use this novel detection method
Charged aerosol detection has the ability to measure any non-volatile and semi-volatile analytes at sub-nanogram, and does not require a chromophore or ionization. This bibliography highlights the breadth and scope of different analysis methods found in the literature.
Download bibliography ›