Go further in your 3D scientific and industrial data analysis
Avizo Software Extensions
Thermo Scientific Avizo Software is a powerful, multifaceted 3D platform for visualizing, manipulating, and understanding scientific and industrial data from computed tomography, microscopy, and many other imaging modalities.
Avizo Software is a configurable solution. Select the extensions that provide the best framework for your 3D data visualization and analysis needs.
Contact us  Request free trial
This extension provides specific support for analyzing fibers, filaments, tunnels, and other networks or tree-like structures. The XFiber extension provides automatic, semi-automatic, and interactive tools to assist segmentation and analysis process. 
Learn more
XFiber
Data courtesy of Rigaku Corporation
High-performance mesher allows for the generation of high-quality 3D tetrahedral mesh for FEA/CFD simulation.This extension is also dedicated to advanced post-processing of simulation data, ranging from flow to thermal, and stress data.
Learn more
XWind
Data courtesy of Visible Cement Dataset
This extension allows for characterization and modeling of a set of connected areas, with spheres and lines representing those areas and their connections.
Learn more
XPoreNetworkModeling
Data courtesy of Shimadzu Corporation (inspeXio SMX-225CT FPD HR)
These extensions provide numerical simulation capabilities to calculate absolute permeability, thermal conductivity, electrical resistivity, and molecular diffusivity.
XLab
Courtesy of Dominique Bernard ICMCB-CNRS
This extension allows you to compute the precise internal displacement and strain maps from 3D images of objects acquired during a deformation process. 
Learn more
XDigitalVolumeCorrelation
Courtesy of Dr Gianluca Tozzi at University of Portsmouth
Manage and visualize very large amounts of volume data of up to several terabytes. Go far beyond the limit of the available system memory.
The multiresolution technique used in XLVolume extension enables interactive visualization and navigation through large data sets. 
Learn more
XLVolume
XXL CT of a complete car. Courtesy of Fraunhofer EZRT
Conveniently load 3D or 4D+ data from over 150 file formats (powered by Bio-Formats, in cooperation with Glencoe Software). 
Learn more
XBioFormats
This extension combines Avizo Software's strong capabilities for 3D data visualization, such as hardware-accelerated volume rendering, with specific tools for molecular visualization and analysis, such as molecular surfaces, sequence alignment, configuration density computation and molecule trajectory animation.
XMolecular
Courtesy of Dr Gianluca Tozzi at University of Portsmouth
Create new custom components for Avizo Software, such as file readers and writers, computation modules, and even new visualization modules, using the C++ programming language.
Use Avizo Software's advanced data visualization and analysis features on immersive VR systems or tiled screens configurations.
XScreen