Focused Ion Beam (FIB) systems replace the low-mass electrons of the SEM and TEM with high-mass ions in order to mill (or sputter) the specimen’s surface, making it possible to produce minute components or to remove unwanted material. Our FIBs are well suited to a wide range of process control, failure analysis and materials research applications.
The Thermo Scientific Aquilos Cryo-FIB is the first cryo-DualBeam (focused ion beam/scanning electron microscope) system dedicated to preparation of frozen, thin lamella samples from biological specimens for high-resolution tomographic imaging in a cryo-transmission electron microscope (cryo-TEM). Cryo-electron tomography’s ability to visualize structures in their native context allows researchers to observe functional relationships and interactions with other components in the cellular environment. This technique promises to become an important tool for scientists seeking a better understanding of living systems at the molecular level.
Available for: Life Sciences Research
The Thermo Scientific V400ACE Focused Ion Beam (FIB) system is specifically designed to meet the challenges of advanced designs and processes: smaller geometries, higher circuit densities, exotic materials, and complex interconnect structures.
Available for: Semiconductors Research
The Thermo Scientific OptiFIB Taipan System is a robust focused ion beam circuit edit system optimized for the 10nm process node. Its highest imaging and milling resolution, highest beam placement accuracy, and low system drift combine to result in better success rates and higher productivity. The OptiFIB Taipan System also features a unique portfolio of circuit edit-centric chemistries and software automation for improved usability.
Available for: Semiconductors Research