The most versatile micro computed-tomography (microCT) solution for quantitative analysis
Versatile micro computed-tomography instruments provide solutions for quantitative analysis for a variety of research applications. Unique helical scanning and iterative reconstruction technology produce unsurpassed image fidelity and deliver the highest signal-to-noise ratio compared to traditional circular scanning technology.
Thermo Scientific HeliScan brings a new era of microCT to Materials Science by leveraging advanced helical scanning and iterative reconstruction technology to produce unsurpassed image fidelity. As part of a multi-scale imaging solution, HeliScan enables scientists to gain valuable insight from internal structures to explore and validate a wide range of material properties.
HeliScan offers image accuracy and resolution that helps geologists, core analysts, petrophysicists and reservoir engineers learn about the microstructural context of their core samples. Bridging the gap between whole core CT and SEM datasets, HeliScan is the ideal choice for visualization of experiments, quantitative property assessments, and high resolution digital archiving of core plug samples.