On this page:
Importance of methane detection
Methane emissions have become a focal point in global efforts to combat climate change due to their significant impact on global warming. Recent regulations have been implemented to curb these emissions, necessitating the use of advanced detection technologies. Discover key regulatory requirements and explore leading detection tools for accurate and effective methane monitoring.
Methane emissions regulations
Governments and environmental agencies worldwide are introducing stringent regulations to limit methane emissions from various industries. Key regulations include:
  • EPA Methane Rule (US): The Environmental Protection Agency (EPA) has set standards for methane emissions from new, modified, and reconstructed sources in the oil and gas industry. This includes requirements for leak detection and repair (LDAR) programs.
  • European Union Methane Strategy: The EU has adopted a comprehensive strategy to reduce methane emissions across the energy, agriculture, and waste sectors.
  • Global Methane Pledge: Over 100 countries have committed to reducing global methane emissions by at least 30% from 2020 levels by 2030.
Advanced methane detection technologies
To comply with methane emissions regulations, industries must utilize reliable and accurate methane detection tools. Below are some of the leading products available:
TVA2020 Toxic Vapor Analyzer
The Thermo Scientific TVA2020 Toxic Vapor Analyzer is designed for the detection of methane and other volatile organic compounds (VOCs). It combines Flame Ionization Detection (FID) and Photoionization Detection (PID) technologies for comprehensive monitoring.
Key features:
  • Dual detection technology (FID and PID)
  • Portable and lightweight
  • Data logging capabilities
  • Meets EPA Method 21 requirements
MIC-6 Multi Instrument Calibrator
The Thermo Scientific MIC-6 Multi Instrument Calibrator is a portable flame ionization detector that offers high sensitivity for methane detection. It is ideal for field use in various environmental monitoring applications.
Key features:
  • High sensitivity and fast response
  • Lightweight and easy to use
  • Long battery life for extended field operations
Direct methane & non-methane hydrocarbon analyzer Model 55i
The Thermo Scientific Model 55i Methane & Non-Methane Analyzer is a back-flush gas chromatography (GC) system is designed for automated measurement of methane and non-methane hydrocarbons.
Key features:
  • Real time correction of THC readings 
  • Automatic flame sensing and ignition 
  • Automatic calibration and span check
Opgal™ EyeCGas™ Multi Optical Gas Imaging Camera
The Opgal EyeCGas Multi Optical Gas Imaging (OGI) Camera is an advanced optical gas imaging tool that allows for the visualization of methane and other gases. It is essential for quick and effective leak detection in various industrial settings.
Key features:
  • Real-time gas visualization
  • Detects multiple gases simultaneously
  • Rugged and suitable for harsh environments
Opgal EyeCGas CO Optical Gas Imaging Camera for Carbon Monoxide 
Detect dangerous CO leaks instantly with Opgal EyeCGas CO, a rugged, intrinsically safe OGI camera built for safety, speed, and reliability in industrial settings.
  • Optimized CO leak detection
  • Thermographic imaging
  • Rugged, lightweight and intrinsically safe
Opgal EyeCGas CO2 Optical Gas Imaging Camera for Carbon Dioxide 
Pinpoint tiny CO2 leaks fast with Opgal EyeCGas CO2, a rugged, intrinsically safe OGI camera built for safety, accuracy, and reliability in harsh industrial environments. 
  • Detection of CO2 leaks in small concentrations
  • Thermographic imaging
  • Rugged, lightweight and intrinsically safe
Opgal EyeCGas 2.0 Long Range Optical Gas Imaging Camera 
This fast, rugged Opgal EyeCGas long-range gas imaging camera detects methane, CO, and VOC leaks in harsh environments, enhancing safety and reducing downtime effectively. 
  • Detects methane, CO2, and over 400 VOCs with high sensitivity
  • Long-range telephoto performance
  • Rugged and intrinsically safe
Conclusion
Adhering to methane emissions regulations is crucial for environmental protection and regulatory compliance. Utilizing advanced detection technologies, such as those offered by Thermo Fisher Scientific, helps to ensure accurate monitoring and effective mitigation of methane emissions. By investing in reliable detection tools, industries can significantly reduce their environmental impact and contribute to global efforts in combating climate change.
Contact us
For more information on methane emissions regulations and to find the right detection tools for your needs, please contact us.
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability.