Search Thermo Fisher Scientific
Rely on us for end-to-end solutions that guarantee productivity at every step of the workflow.
Test for the most common spoilage microorganism species of interest, with over 100 strains, including yeast and mold, Brettanomyces spp., preservative-resistant yeast, lactic acid bacteria, and acetic acid bacteria, using the SureTect Beverage Spoilage Multiplex PCR Assay. Developed in partnership with a major global beverage manufacturer to meet the industry’s needs, the assay is an optimized solution combining speed and performance in an easy-to-use and cost-effective platform.
Thermo Scientific Nalgene Analytical Filter Funnels and Cellulose Membrane Filters effectively recover microorganisms.
Dark color membranes inproves visualization of colonies
Flexible formats - single wrapped, dispenser filter and funnel
Wide range of filter and media certified tests available
Filter tested on high quality and ISO conform media
Thermo Scientific Remel and Oxoid Culture Media offers a broad range of dehydrated and prepared products in a variety of sizes and formats. Learn about our Dehydrated Culture Media (DCM) Convenience Formats.
Formulation optimized for recovery and differentiation of organisms
Global manufacturing network offers reliable product supply
ISO certified manufacturing ensures consistent media performance
Chromogenic media supports rapid differentiation and easy identification
The Thermo Scientific SureTect PCR System offers an easy-to-use workflow for rapid and accurate detection of all common food pathogens and some quality organisms.
Ready to use reagents and ergonomically designed tools for easy handling
Analyze multiple pathogen detection kits simultaneously
Color coded plates and reagents to keep you on track
Smart instrumentation for automated lysis, test set up and result interpretation
Thermo Scientific Culti-Loops Quality Control Organisms offer stabilized, preserved, viable microorganisms for simple and convenient performance testing.
Extensive range of ATCC* and NCTC organisms
Loop format for lower risk of contamination and consistent results
Gel preserved organisms for faster recovery
Peel-off transferable labels for improved traceability
*The ATCC Licensed Derivative® Emblem, the ATCC Licensed Derivative® word mark and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc., and not ATCC.
For Research Use Only. Not for use in diagnostic procedures.