Improving food development, from ingredients to consumables, with rheology and extrusion solutions
Developing sustainable food ensures that the growing global population has access to sufficient, high-quality, affordable, and nutritious food while minimizing environmental impact and meeting consumer expectations for taste and texture. Key trends in sustainability include plant-based proteins and natural, functional ingredients that support health.
Extrusion technology is a promising platform for food, feed, or nutritional development, recognized for its energy efficiency and environmental friendliness. It offers a cost-effective, continuous production process with precise control to maintain high product quality.
Rheological characterization optimizes the mechanical properties and sensory perceptions of food products to meet high consumer expectations regarding taste and texture, and to simulate processing conditions. Rheology can also help estimate the stability and shelf life of a complex food formulation.
The stability of a system like food emulsions or food dispersions can be predicted by measuring the formulation’s viscoelastic properties. Viscosity measures how easily a liquid flows, and yield stress measurements provide information about the energy needed to overcome elasticity in semi-solid formulations. 
The flexibility of rheometers, with a wide range of application-specific measuring cells for powder rheology, tribology, texture, and interfacial analysis, or even combined optical methods, allows for comprehensive product characterization. This holistic approach ensures that food products not only meet nutritional needs and environmental goals but also deliver the desired sensory experiences that consumers demand.
Understand flow behavior of food products, from processing to mouthfeel
Understand how to meet technical requirements and customer expectations for texture and mouthfeel, as well as for processing of a variety of foods.
Introduction on high and low moisture extrusion of plant proteins
Learn how twin-screw compounders are a flexible solution for the development of plant-based proteins. 
Confectionary
The confectionery industry, which focuses on products made from sugar or sugar substitutes, is diverse, including chocolates, candy bars, jellies, gums, hard and chewy candies, and other sweets. Factors such as flow properties, ingredient interactions, response to deformation, temperature changes, and shelf stability are investigated with rheological characterization. Innovative technologies like co-rotating twin-screw extrusion technology offer the possibility of continuous manufacturing of confectioneries. This method simplifies the process and product development by minimizing the number of production steps, resulting in significant time and lab space savings. It allows precise control over the molecular alterations of proteins, sugars, starches, and fats, ensuring high-quality confectionery products.
Chocolate
The flow behavior of some foods, specifically the viscous behavior of molten chocolate, affects the surface appearance and mouthfeel properties of the final confection. Viscosity and yield stress also play important roles in chocolate manufacturing, affecting transport, filling, dipping, coating, dosing, and storage operations. 
Learn more about chocolate testing:
Expanded food products
Breakfast cereals, snacks, and texturized vegetable proteins are common expanded food products that are produced using extrusion technology. The extrusion process serves to impart a unique puffy, crunchy texture into these products with a variety of shapes ranging from balls, rings, to curls, and many more. Depending on the plant source, breakfast cereals and snacks can be made from ingredients such as rice, corn, and potato. Protein-rich ingredients are applied to manufacture texturized vegetable proteins that serve as meat replacers. Even though extrusion is an established production process, new ingredients that are selected to improve the nutritional value, and ecological footprint of these products, require constant adaptation of process parameters to maintain desired texture and mouthfeel.
Learn more about expanded food products:
Diary, spreads and sauces
Consumer acceptance of dairy foods depends on their textural characteristics from usage like spreadability up to sensory perception such as texture and mouthfeel. Adapting to changing market trends, such as plant-based alternatives and lactose-free options, adds complexity. Consequently, to enhance consumer acceptance, both formulation and production processes must be adapted to achieve the desired flow properties, texture, and mouthfeel.
Learn more about:
Encapsulation of flavors and active ingredients
Flavors and active ingredients are sensitive and expensive additives. Encapsulation serves multiple purposes, such as protecting against oxidation, preventing flavor degradation, and enhancing product handling. Utilizing twin-screw extruders for encapsulating flavors into a matrix provides several benefits, including gentle processing conditions that minimize the risk of product degradation. Additionally, various downstream equipment options allow for the customization of end products into different shapes, such as pelletized powder or chill-rolled flakes.
Plant based meat and dairy alternatives
Plant-based ingredients can be formulated to mimic the texture, taste, appearance, and nutritional value of animal meats. These imitation meat products are rising in popularity at grocery stores for home use and in restaurants everywhere. Twin-screw extrusion enables consistent production of meat analogs to mimic beef, chicken, and seafood.
Plant based dairy alternatives
Plant-based dairy alternatives are increasingly popular for their health, sustainability, and ethical benefits. Therefore, the development of alternative protein formulations and valorization of food side streams are key focuses. Sustainable food must meet consumer expectations for taste and texture. Rheological characterization optimizes mechanical properties and sensory perceptions to meet these expectations and simulate processing conditions. The high flexibility of rheometers, with various application-specific measuring cells for tribology, texture, and interfacial analysis, allows comprehensive product characterization.
Learn more about plant-based dairy alternatives:
Starch
Create better starch products by leveraging the unique properties of starch from various plant sources. Understand viscosity, texture, and stability for a wide range of applications. Native starch has a grain-like structure with small crystalline particles. Cooking starch in water breaks this structure, creating a starch solution. The viscosity and texture of this solution or paste, as well as its storage stability, vary based on the starch source and pre-treatment. During cooking, viscosity peaks due to starch crystal swelling and then decreases as crystalline domains break.
Read more Investigation of food structures
In order to obtain information about the reasons for certain changes in rheological properties, modern rheometers offer the possibility of combined methods, e.g. with the coupling of an optical microscope.
Webinar: How extrusion conditions influence the properties of starch compounds
Watch this webinar to see the benefits of adeptly designing a starch matrix by twin-screw extrusion.  Learn how to influence the texture, stability and further processability of the final product via extrusion.
For Research Use Only. Not for use in diagnostic procedures.