Which X-ray Detection system is right for you?
Select an application below
Retail Packs
Retail packs
Entry-Level X-ray System NextGuard™ C330
Designed for a wide variety of food applications, NextGuard systems offer enhanced capabilities to inspect packaged products for missing pieces or components, under and over-fills, and other quality problems with easily customizable vision software. Standalone rejection conveyors for use after the x-ray inspection system are also available.
Max inspectable area13.4 in. wi. x 7.3 in. ht.
Washdown ratingIP65 with AC
Typical sensitivity0.8–1.5 mm SST, 3.5–5 mm glass
Max product weight22 lbs
X-ray beam orientationVertical, bottom up, no blind spots
Max x-ray power160W
NextGuard™ C330 PRO
The Pro model offers a wide range of single-belt configurations with integrated rejection systems, shielding, heavy-duty motor/rollers and stainless steel flange bearings for more demanding large packages and wet applications. The conveyor can handle up to 5x more weight and can be extended in length up to 3.5m. The Pro can also utilize up to three AC variable frequency drives, making complex multi-conveyor custom configurations possible.
Max inspectable area13.4 in. wi. x 7.3 in. ht.
Washdown ratingIP65 with AC
Typical sensitivity0.8–1.5 mm SST, 3.5–5 mm glass
Max product weight100 lbs
X-ray beam orientationVertical, bottom up, no blind spots
Max x-ray power160W
High Performance Xpert™ Conveyor
These high performance vertical X-ray beam (top down) systems perform X-ray contaminant detection and product inspection for bags, boxes and other flat packages, quickly identifying contaminants and quality defects that other systems miss. These models are upgradable for harsh washdown environments and come with many options to extend conveyor length.
Max inspectable areaFrom 14.2 in. wi. X 7.1 in. ht . to 29.9 in. wi. X 10.6 in. ht.
Washdown ratingIP65 standard, IP69K optional
Typical sensitivity0.8-2 mm SST, 3.5-7 mm glass
Max product weightFrom 100 to 132 lbs
X-ray beam orientationVertical, top down
Max x-ray power160 - 480 W
Cases and Large Bags Packaging
Cases, large bags
Large Aperature NextGuard™ C500
Designed for a wide variety of food applications, NextGuard C500 systems offer enhanced capabilities to inspect larger packages to small cases with the same benefits of the C330 model.
Max inspectable area19.7 in. wi. x 9.8 in. ht.
Washdown ratingIP65
Typical sensitivity1.5–2 mm SST, 4–7 mm glass
Max product weight25 lbs
X-ray beam orientationVertical, bottom up, no blind spots
Max x-ray power200W
NextGuard™ C500 PRO
The Pro model offers a wide range of single-belt configurations with integrated rejection systems, shielding, heavy-duty motor/rollers and stainless steel flange bearings for more demanding large packages and wet applications. The conveyor can handle up to 5x more weight and can be extended in length up to 3.5m. The Pro can also utilize up to three AC variable-frequency drives, making complex multi-conveyor custom configurations possible.
Max inspectable area19.7 in. wi. x 9.8 in. ht.
Washdown ratingIP65
Typical sensitivity1.5–2 mm SST, 4–7 mm glass
Max product weight100 lbs
X-ray beam orientationVertical, bottom up, no blind spots
Max x-ray power200W
High Performance Xpert™ Conveyor
These high performance vertical X-ray beam (top down) systems perform X-ray contaminant detection and product inspection for bags, boxes and other flat packages, quickly identifying contaminants and quality defects that other systems miss. These models are upgradable for harsh washdown environments and come with many options to extend conveyor length.
Max inspectable areaFrom 14.2 in. wi. X 7.1 in. ht . to 29.9 in. wi. X 10.6 in. ht.
Washdown ratingIP65 standard, IP69K optional
Typical sensitivity0.8-2 mm SST, 3.5-7 mm glass
Max product weightFrom 100 to 132 lbs
X-ray beam orientationVertical, top down
Max x-ray power160 - 480 W
High Performance Xpert™ Heavy Duty Conveyor
High-power, robust solutions that easily detect a wide range of foreign materials in larger cases, bags and trays. Three X-ray inspection widths are available to optimize performance in your food production environment.
Max inspectable areaFrom 14.9 in. wi. X 9.8 in. ht. to 30.3 in. wi. X 11.8 in. ht.
Washdown ratingIP65 standard, IP69K optional
Typical sensitivity1.5–2.5 mm SST, 4–7 mm glass
Max product weightFrom 66 to 88 lbs
X-ray beam orientationVertical, top down
Max x-ray power900W
Bulk Packaging
Bulk
High Performance Xpert Bulk Conveyor
The high performance X-ray system is designed for inspection of free flowing granulated materials. This system finds contaminants such as metal, rock, plastic and glass with removal of product from the line prior to further processing, providing instant cost savings. Xpert systems include powerful software to eliminate false rejects. Multilane inspection and rejection minimizes scrap and rework in this application.
Max inspection width11.3 to 18.7 in.
Washdown ratingIP65
Typical sensitivity1-2mm SST, 3-5mm glass
X-ray beam orientationVertical, top down
Max x-ray power100W - 160W
Plastic Bottles and Cans
Plastic bottles/cans
Xpert S400 Sideshoot
Designed for inspection of tall upright cans, bottles, jars and packages; the single beam Xpert S400 side shoot inspection system provides superior detection of foreign materials. An existing conveyor can be routed through it seamlessly. A chicane configuration is also available for small diameter packages that can't pass through the radiation shielding curtains without tipping over.
Max inspectable area7" W x 10.8" H
Washdown ratingIP65 (IP69K is optional)
Typical sensitivityIn body: 1.5-2.5mm SST & 3-5mm glass
In bottom: 2.5-4mm SST & 5-7mm glass
Shape & thickness of container affects detection capabilities. Testing is recommended to fully understand performance in your product.
Max product weightN/A external conveyor
X-ray beam orientationHorizontal, back to front
Max x-ray power480W
CMD SchemaApp code