Use this comparison chart to see which system is right for you
Still have questions? We're here to help!
NextGuard C330 | NextGuard C500 | NextGuard Pro | Xpert Conveyor | Xpert Bulk | Xpert Sideshoot | |
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
|
Max inspectable area | 13.4 in. wi. x 7.3 in. ht. | 19.7 in. wi. x 9.8 in. ht. | 19.7 in. wi. x 9.8 in. ht. | 15.7 in. wi. x 10 in. ht. | 19.4 in. wi. x2 in. ht. | 7 in. wi. x10.8 in. ht. |
Package Type | ||||||
Cartons/boxes | ✓ | ✓ | ✓ | ✓ | ||
Pouches/bags | ✓ | ✓ | ✓ | ✓ | ||
Bulk flow products |
|
|
|
✓ | ||
Plastic bottles and cans |
|
|
|
|
|
✓ |
Washdown rating | IP65 with AC | IP65 | IP65, stainless steel bearings and gear motor | IP65 standard, IP69K optional | IP65 | IP65 |
Typical sensitivity | 0.8–1.5 mm SST, 2–5 mm glass | 1.5–2 mm SST, 4–7 mm glass | Same as standard models | 1–2 mm SST, 4–7 mm glass | 1–2 mm SST, 3–5 mm glass | 1–2 mm SST, 3–5 mm glass |
Max X-ray power | 160W | 200W | Same as standard models | 315W | 315W | 315W |
Relative price | $ | $$ | $$$ | $$$$ | $$$$ | $$$$ |
Max product weight | 13 lbs | 24 lbs | 110 lbs | 88 lbs | 88 lbs | N/A external conveyor |
X-ray beam orientation | Vertical, bottom up, no blind spots | Vertical, bottom up, no blind spots | Vertical, bottom up, no blind spots | Vertical, top down | Vertical, top down | Horizontal, back to front |
Belt speed range | 10–120 m/min | 5–50 m/min | 5–120 m/min | 5–60 m/min | 5–60 m/min | 5–80 m/min |
Single conveyor with reject integrated | ✓ | ✓ | ✓ | N/A external conveyor | ||
Multi-lane operation | ✓ | ✓ | ✓ | ✓ | ✓ | |
Remote technical support | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |