In Food Safety and Food Testing, Never Sacrifice Product Quality for a Fast Release.
Increase your product quality and decrease your time to market with Life Technologies E. coli O157:H7 testing solutions.
  • Release your product fast and minimize your inventory holdings - accurate and specific detection of E. coli O157:H7 in as little as 8 hours.
  • Reduce risk of false positives  - highly specific TaqMan® chemistry primer/probe designs based on whole-genome sequencing limits cross-reactivity with related E. coli and other non-E. coli organisms.
  • Confidence in your results - external approvals and validation: MicroSEQ® E. coli O157:H7 AOAC & AFNOR validation, Dynabeads® E. coli O157:H7 recommended in FDA BAM method.
Step-by-Step Guide to E. coli Testing Solutions
Universal spin-column-based format for use with all food sample types. Bead-based chemistry for the high-quality purification of DNA from a variety of food samples. Magnetic particle processing instrument for the rapid processing of up to 96 samples.
Complete, validated assays dried down into a single reaction bead. Simply add your purified DNA and get your results in this easy-to-use format. Premier qPCR instrumentation from a leader in molecular analysis equipment. Utilize fast qPCR chemistries to process 96 samples typically in under an hour.
RapidFinder™ Express Software works with your 7500 Fast Real-Time PCR Instrument to provide automated pathogen detection calls and report generation. Minimizes the chances for user error or operator bias.
Specifically enrich your sample reducing the background organism making detection easier and more specific.Automated magnetic processing system for processing of up to 15 samples in parallel. Automation brings ease-of-use and consistency to your IMS cell isolation.