Through high-affinity antibodies attached to the surface of magnetic Dynabeads® - known as immunomagnetic separation (IMS) technology - a valuable means to isolate specific biological entities from complex matrices is provided for your work!  Their use in sample preparation greatly improves the speed, sensitivity, and reproducibility of microbial analysis.  Dynabeads® IMS water products have been designed for both manual and automated separation of the specific target organism.
Invitrogen has entered into a strategic collaboration with IDEXX Laboratories, Inc. for the distribution and development of new products for the water testing industry. Please contact IDEXX for the purchase of Invitrogen’s Dynabeads® anti-Cryptosporidium kits and Dynabeads® GC-Combo kits. For more information, visit the IDEXX website at www.idexx.com/water or phone the IDEXX office at:
USA - 1-800-321-0207 or 207-556-4496
Europe - +800-433-991-11 or +44-1638-676800
Product Applications
Dynabeads® anti-Cryptosporidium Kit For the selective enrichment of Cryptosporidium oocysts directly from water sample concentrates. Performance tested: Validated IMS in the US Environment Protection Agency’s Method 1622 and by UK DETR Water Supply Regulation 1999 SI No 1524.
Dynabeads® GC-Combo For the selective enrichment of Giardia cysts and Cryptosporidium oocysts directly from water sample concentrates. Performance tested: Validated IMS in the US Environment Protection Agency’s Method 1623
Dynal L10 tubes Flat-sided reusable glass tubes necessary for optimal recovery of cysts / oocysts during magnetic separation with the Dynal MPC-1 when using Dynabeads® anti-Cryptosporidium or Dynabeads® GC-Combo kits.
Dynal Spot-On Treated single 9mm well, glass slides for microscopic analysis of oocysts and cysts. This product is used in combination with the Dynabeads® anti-Cryptosporidium and Dynabeads® GC-Combo kits.
For research use only. Not intended for any animal or human therapeutic or diagnostic use.