RH-system
Automated STR-based human identification
Product description
Part number category
Part number
Price
RapidHIT System Instrument
Instrument
100010RH
Request a Quote
RapidHIT GlobalFiler Express 3-Sample Cartridge Kit (12 Samples per Kit)
Consumables
400084RH
Request a Quote
RapidHIT GlobalFiler Express 7-Sample Cartridge Kit (28 Samples per Kit)
Consumables
400086RH
Request a Quote
RapidHIT GlobalFiler Express 8-Sample Cartridge Kit(32 Samples pr Kit)
Consumables
400106RH
Request a Quote
RapidHIT NGM SElect Express 7-Sample Cartridge Kit(28 Samples per Kit)
Consumables
400090RH
Request a Quote
RapidHIT NGM SElect Express 3-Sample Cartridge Kit Plus (12 Samples per Kit)
Consumables
400102RH
Request a Quote
RapidHIT NGM SElect Express 8-Sample Cartridge Kit(32 Samples per Kit)
Consumables
400105RH
Request a Quote
RapidHIT, Storage Cartridge Kit (10 uses)
Consumables
400061RH
Request a Quote
RapidHIT Blank Cartridge Kit (4 pack; 20 runs per cartridge)
Consumables
400087RH
Request a Quote
Description:
The IntegenX by Thermo Fisher Scientific RapidHIT System is the first fully-automated, sample-to-answer system for STR-based human identification. RapidHIT is available to forensic scientists in booking stations, laboratories, and in the field home or abroad.
The self-contained human identification system produces standardized DNA profiles from buccal swabs and other human samples in under two hours. Requiring only three minutes of hands-on time to generate up to eight DNA profiles in one run, the easy-to-use system fits into any forensic workflow. The RapidHIT system uses NDIS-approved Applied Biosystems GlobalFiler Express chemistry, and the output from the system is compatible with local, state and national human identification databases.
  • >90% full profile and CODIS success rate
  • 1 to 8 samples in one run
  • Process any swab type and certain crime scene samples directly in sample cartridge
  • Requires only 3 minutes of hands-on time, less than 2 hours time to results
Specifications:
Dimensions and weight
73 x 71 x 48 cm (28.5 x 28 x 18 in)
81.5 kg (180 lbs)
Performance
Up to 8 samples per run
Processing time under 2 hours
Allelic ladder control
Applied Biosystems GlobalFiler Express–24 markers including expanded core CODIS loci
Applied Biosystems AmpFLSTR NGM SElect Express–17 markers
Certificates
Manufactured under ISO 9001
CE and TUV
Operating environment 18–30ºC (59–86ºF)
15–75% relative humidity
Elevation up to 2000 m (6500 ft)
100–240 VAC, 50/60 Hz, 120V, 400W (peak at 600W), recommended 1000 VA uninterruptible power supply
For Forensics, Human Identification or Paternity/Kinship Use Only. Not for use in diagnostic or therapeutic applications.