Each Quantifiler® Duo kit includes all reagents necessary for the amplification, detection, and quantification of 400 samples in 25 ul reactions using the Applied Biosystems 7500 Real-Time PCR System.
| |||||||||||||||
Search Thermo Fisher Scientific
Each Quantifiler® Duo kit includes all reagents necessary for the amplification, detection, and quantification of 400 samples in 25 ul reactions using the Applied Biosystems 7500 Real-Time PCR System.
| |||||||||||||||