4N6FLOQSwabs genetics devices utilize proprietary flock technology to maximize DNA collection and elution efficiency. The perpendicular nylon fibers act like a soft brush that facilitates improved collection of cellular material. Unlike traditional fiber-wound swabs, 4N6FLOQSwabs devices have no internal absorbent core to disperse and entrap the specimen—the sample stays close to the surface for faster and more efficient elution.
Key features
- Validated for direct PCR amplification: reference samples collected with 4N6FLOQSwabs reference collection devices can be easily processed using Applied Biosystems Prep-n-Go Buffer, eliminating the need for extraction and quantification
- Easy processing with prescored breaking point for breaking of the swab into tube
- ISO 18385–compliant and certified free of DNase, RNase, and amplifiable human DNA
- Specially designed for buccal sample collection and available in different tubing configurations.
- Customized solutions for manufacturing kits for the sample collection workflow that meet the challenging, unique needs of buccal sample collection.
- Improved chain of custody with peel-off barcode feature.
Featured product
4N6FLOQSwabs (Cat. #4479433) Regular Tip with plastic tubing is a versatile collection device with a solid, molded plastic applicator shaft and a regular-sized tip that can vary in size and shape.
Workflow
Ordering information
1Typical workflow for 24 samples using the latest Applied Biosystems direct PCR amplification kits, genetic analyzers, and expert system analysis software. *To order please contact your local Life Technologies sales representative.
For Forensic or Paternity use only. 4N6FLOQSwabs is a trademark of Copan Flock Technologies.