Identifiler Direct now just "Punch & Go"
Now you can simply punch a disc from a standard FTA® card and go straight to amplification. The AmpFℓSTR® Identifiler® Direct PCR Amplification Kit amplifies the 16 loci included in the Identifiler® kit from single source samples spotted on FTA® cards, without the need to perform any DNA extraction or purification. By eliminating these tedious pre-amplification steps, the Identifiler® Direct kit simplifies automation requirements and expands throughput capabilities, while maintaining high profile quality and maximizing quality assurance for the entire process.
Key Features
  • Enables the direct amplification of blood and buccal samples without the need for extraction or purification
  • Facilitates high throughput & automated sample processing
  • Minimizes the risk of contamination and procedural errors by reducing the number of processing steps
  • Helps accelerate time to result with minimal reduction in data quality
  • Minimizes extraction⁄purification costs and helps reduce the cost of labor and consumables
Learn more
How it works & Comparison to Other Methods