Turn samples Into answers in less than 2 hours
This breakthrough chemistry enables up to 5 times faster amplification time than previous-generation kits:
  • GlobalFiler™ Express Kit for single source samples: ~40 minute amplification
  • GlobalFiler™ Kit for casework samples: ~80 minute amplification
Typical workflow for up to 48 samples using Applied Biosystems® direct PCR amplification kits, genetic analyzers and expert system analysis software.