4N6FLOQSwabs™ DNA Collection System
4N6FLOQSwabs™ devices utilize proprietary flocked-swab technology to maximize DNA collection and elution efficiency. The perpendicular Nylon® fibers act like a soft brush, which improves collection of cellular material. Unlike traditional fiber wound swabs, 4N6FLOQSwabs™ devices have no internal absorbent core to disperse and entrap the specimen—the sample stays close to the surface for faster and more efficient elution.
Key features
- Designed for rapid absorption and superior sample release, enabling increased assay sensitivity and performance for both reference and casework samples
- Pre-scored breaking point allows for easy breaking of the swab into tubes for processing
- Certified free of DNase, RNase, and amplifiable human DNA; uses only selected non-inhibitory components
- Available in a variety of ergonomic designs, configurations, and customizable collection kits
- ETO-treated (Ethylene Oxide)
Designed specifically for crime scene sample collection:
The unique 4N6FLOQSwabs™ DNA Collection System Crime Scene product line is specifically designed for the collection of sweat, semen, blood stains, skin, and environmental trace evidence found at the crime scene.
- Optimized for challenging samples—rapid absorption capability and superior sample release is especially valuable with limited or trace amounts of DNA
- Prevents microbial contamination—swabs contain antimicrobial agents for improved sample integrity
- Sample drying not needed—effective antimicrobial action allows the sample to be transferred into a tube or vial without drying and to maintain integrity even in more humid conditions
Figure 1. The complete integrated workflow for efficient evidence sample processing from Life Technologies.
4N6FLOQSwabs™ Devices |
Cat. No. |
Quantity |
4N6FLOQSwabs™ Regular head tip, in peelpouch with Antimicrobial Action |
Case of 100 |
|
4N6FLOQSwabs™ Regular head tip, in plain tube with Antimicrobial Action |
Case of 100 |
|
4N6FLOQSwabs™ Scalpel shape tip, in plain tube with Antimicrobial Action |
Case of 100 |
|
4N6FLOQSwabs™ Regular Mini head tip, in plain tube with Antimicrobial Action |
Case of 100 |
|
4N6FLOQSwabs™ Regular head tip, in plastic short tube with Antimicrobial Action |
Case of 50 |
|
4N6FLOQSwabs™ Dual Swab Regular size tip in plain tube with Antimicrobial Action |
Case of 100 |
|
4N6FLOQSwabs™ Subungual Shape, in plain tube with Antimicrobial Action |
Case of 50 |
|
4N6FLOQSwabs™ Round Shape, in plain tube with Antimicrobial Action |
Case of 50 |
|
4N6FLOQSwabs™ Mini tip, in plain tube with Antimicrobial Action |
Case of 50 |
© 2013 Life Technologies Corporation. For Forensic or Paternity use only. All rights reserved. The trademarks mentioned herein are the property of Life Technologies Corporation and/or its affiliate(s) or their respective owners. 4N6FLOQSwabs is a trademark of Copan Flock Technologies.