4N6FLOQSwabs™ devices utilize proprietary flocked-swab technology to maximize DNA collection and elution efficiency. The perpendicular Nylon® fibers act like a soft brush, which improves collection of cellular material. Unlike traditional fiber wound swabs, 4N6FLOQSwabs™ devices have no internal absorbent core to disperse and entrap the specimen—the sample stays close to the surface for faster and more efficient elution.
Key features
  • Designedfor rapid absorption and superior sample release, enabling increased assay sensitivity and performance for both reference and casework samples
  • Pre-scored breaking point allows for easy breaking of the swab into tubes for processing
  • Certified free of DNase, RNase, and amplifiable human DNA; uses only selected non-inhibitory components
  • Available in a variety of ergonomic designs, configurations, and customizable collection kits
  • ETO-treated (Ethylene Oxide)
Figure 1. An example of a profile generated from a 4N6FLOQSwabs™ device sample using the Prep-n-Go™ Buffer and GlobalFiler™ Express Kit.
Designed specifically for reference sample collection:
The unique 4N6FLOQSwabs™ DNA Collection Reference Sample product line is specifically designed for faster absorption and more efficient sample release as compared to traditional swabs.
  • Validated for direct PCR amplification, therefore single source samples collected with 4N6FLOQSwabs™ devices can be easily processed using the Applied Biosystems® Prep-n-Go™ Buffer, eliminating the need for extraction steps
  • Available in different formats to enhance sample preservation to meet the varied needs of forensic and paternity laboratories
Figure 2. The complete, integrated workflow from Life Technologies for single source-samples using the 4N6FLOQSwabs™ device enables laboratories to go from sample to result in as little as 2 hours.1
4N6FLOQSwabs™ DevicesCat. No. Quantity
4N6FLOQSwabs™ Lollipop Swab in peelpouch 4473997Case of 50
4N6FLOQSwabs™ Regular size tip in peelpouch 4473979Case of 100
4N6FLOQSwabs™ Regular size tip in plain tube 4479433*Case of 100
4N6FLOQSwabs™ Regular size tip in plastic tube with Active Drying System 4479439*Case of 50
4N6FLOQSwabs™ Regular size tip plus 2ml EPPENDORF® cuvette in peelpouch 4479431*Case of 100
4N6FLOQSwabs™ Regular size tip plus 2ml EPPENDORF® cuvette with evaporation duct in peelpouch 4479440*Case of 100
4N6FLOQSwabs™ Dual Swab Regular size tip in plain tube 4479432*Case of 100
4N6FLOQSwabs™ Buccal shape tip in plastic tube with Active Drying System 4479436*Case of 50