The following databases allow DNA labs to search and estimate the Y-STR haplotype frequency across populations for forensic and paternity casework. Yfiler™ and Yfiler™ Plus kits are engineered for >99.99% concordance to facilitate upgrade between the two kits and maximum usage of available Y-STR databases.
 Database Total haplotypes Total Yfiler & Yfiler Plus
searchable haplotypes*
 Notes
Y-STR Haplotype Reference Database (YHRD) v4.0
http://www.yhrd.org
143,044 93,302
Established by Lutz Roewer and Sascha Willuweit (Charité - Universitätsmedizin Berlin), this database is updated continually through user submissions
US Y-STR Database v4.1
http://usystrdatabase.org
35,295 26,683
Funded by the US National Institute of Justice and managed by the National Center for Forensic Science (NCFS)
Using the STR Haplotype Reference Database (YHRD)
GlobalFiler kits and Yfiler kits: For Forensic or Paternity Use Only.
The Quantifiler kits are For Research, Forensic or Paternity Use Only. Not for use in diagnostic procedures.