Meet the highest standards of performance, hygiene and reliability
Pharmaceutical applications demand the highest standards of performance, hygiene and reliability. Thermo Scientific product inspection solutions can detect incomplete packages at high speed through high accuracy dynamic weight check of packed and part-packed goods or detect metallic and non-metallic contamination in both oral solid production or in liquid vial filling. From the consistent accuracy of our pharmaceutical checkweighers, to the sensitivity of our metal detectors for the pharmaceutical industry, to the detection performance of our x-ray inspection systems, our solutions are proven effective and expertly tailored to inspect pharmaceutical packaging.
Created to provide outstanding quality control of package contents, the Versa Rx Pharma checkweigher meets the most stringent requirements for validated production. It can detect the presence or absence of the lightweight descriptive pamphlet that is legally required within cartoned products, and is capable of weighing products from 2 g to 500 g at speeds up to 550 ppm—depending on the size of the pack.
The Global Versa GP checkweigher is versatile and for general purpose use. It scans at line speeds up to 300 packages per minute without sacrificing accuracy and comes with a compact cantilever design for easy height adjustment. It is Ideal for packages up to 3 kilograms (6.6 pounds).
The Global VersaWeigh is a general purpose checkweigher designed to provide superior levels of quality control in the pharmaceutical packaging environment.It is a highly reliable, easy-to-use and easy-to-maintain system that complies with Good Manufacturing Practice (GMP) guidelines.
Our pharmaceutical metal detector is a mobile, versatile and reliable metal detection solution for tablet presses, capsule filling and deduster applications. Achieve cost-effective, industry-leading sensitivity in the most demanding pharmaceutical environments while meeting the U.S. Food and Drug Administration's (FDA) stringent requirements for validated production.
The Thermo Scientific POWERx D165 X-ray inspection system for glass contaminant detection is designed to offer the highest level of quality assurance possible. It features state-of-the-art X-ray sources, detectors and image analysis software.
Our easy-to-use X-ray inspection system provides complete protection from metal, glass, and other dense foreign objects. The Thermo Scientific NextGuard conveyor X-ray detection system offers enhanced capabilities to inspect packaged products for missing pieces or components, under and over-fills, and other quality problems with easily customizable vision software.
Video spotlight
Thermo Scientific product inspection systems for pharmaceutical and biotech applications provide valuable quality and safety checks for pharmaceutical manufacturers. Read these frequently asked questions and answers about our online pharmaceutical product inspection systems.