Use this comparison chart to see which system is right for you
|
RM 100 PL |
RM 110 PL |
RM 200 PL |
RM 210 PL |
Laser TX |
SIPRO |
RM 200 CM |
RM 210 CM |
RM 215 HM |
RM 302 ES |
RM 200 EG |
RM 316 EG |
Sensor |
Isotope |
X-ray |
Isotope |
X-ray |
Optical |
X-ray |
Isotope |
X-ray |
X-ray |
Isotope |
Isotope |
X-ray |
Process lines |
Coating lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Slitting lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Inspection lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Cut-to-length lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Pickling lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Tension leveling |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Cold mills |
Reversing cold mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Tandem cold mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
PLTCM |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Z-mills/ other mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Hot mills |
Multi-stand HM |
|
|
|
|
|
✓* |
|
|
✓ |
|
|
|
Steckel mill |
|
|
|
|
|
✓* |
|
|
✓ |
|
|
|
Plate mill |
Roughing mill |
|
|
|
|
|
|
|
|
|
|
✓ |
✓ |
Shear lines |
|
|
|
|
|
|
|
|
|
|
✓ |
✓ |
*Preferred offering |
|
RM 210 AF |
RM 210 AS |
RM 210 AH |
SIPRO |
Sensor technology |
X-ray |
X-ray |
X-ray |
X-ray |
Process lines |
Coating lines |
|
✓ |
|
|
Slitting lines |
✓ |
✓ |
|
|
Inspection lines |
|
✓ |
|
|
Cut-to-length lines |
|
✓ |
|
|
Pickling lines |
|
✓ |
|
|
Tension leveling |
|
✓ |
|
|
Cold mills |
Reversing cold mills |
✓ |
✓ |
|
|
Tandem cold mills |
✓ |
✓ |
|
|
Pickling line/ tandem cold mill |
✓ |
✓ |
|
|
Foil mills |
✓ |
|
|
|
Hot mills |
Multi-stand HM |
|
|
✓ |
✓* |
Plate mill |
Roughing mill |
|
|
✓ |
|
Cut-to-length lines |
|
|
✓ |
|
* Preferred offering |
|
RM 100 PL |
RM 110 PL |
RM 200 PL |
RM 210 PL |
Laser TX |
SIPRO |
RM 200 CM |
RM 210 CM |
RM 215 HM |
RM 302 ES |
RM 200 EG |
RM 316 EG |
Sensor |
Isotope |
X-ray |
Isotope |
X-ray |
Optical |
X-ray |
Isotope |
X-ray |
X-ray |
Isotope |
Isotope |
X-ray |
Process lines |
Coating lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Slitting lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Inspection lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Cut-to-length lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Pickling lines |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Tension leveling |
✓ |
✓ |
✓ |
✓ |
✓* |
|
|
|
|
✓ |
|
|
Cold mills |
Reversing cold mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Tandem cold mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
PLTCM |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Z-mills/ other mills |
|
|
|
|
|
|
✓ |
✓* |
|
|
|
|
Hot mills |
Multi-stand HM |
|
|
|
|
|
✓* |
|
|
✓ |
|
|
|
Steckel mill |
|
|
|
|
|
✓* |
|
|
✓ |
|
|
|
Plate mill |
Roughing mill |
|
|
|
|
|
|
|
|
|
|
✓ |
✓ |
Shear lines |
|
|
|
|
|
|
|
|
|
|
✓ |
✓ |
*Preferred offering |
|
RM 210 AF |
RM 210 AS |
RM 210 AH |
SIPRO |
Sensor technology |
X-ray |
X-ray |
X-ray |
X-ray |
Process lines |
Coating lines |
|
✓ |
|
|
Slitting lines |
✓ |
✓ |
|
|
Inspection lines |
|
✓ |
|
|
Cut-to-length lines |
|
✓ |
|
|
Pickling lines |
|
✓ |
|
|
Tension leveling |
|
✓ |
|
|
Cold mills |
Reversing cold mills |
✓ |
✓ |
|
|
Tandem cold mills |
✓ |
✓ |
|
|
Pickling line/ tandem cold mill |
✓ |
✓ |
|
|
Foil mills |
✓ |
|
|
|
Hot mills |
Multi-stand HM |
|
|
✓ |
✓* |
Plate mill |
Roughing mill |
|
|
✓ |
|
Cut-to-length lines |
|
|
✓ |
|
* Preferred offering |
Sensor/system compatibility chart
|
Isotope |
Isotope |
X-ray |
X-ray |
X-ray |
X-ray |
Optical |
|
Am-241 |
Cs-137 |
<50 kV |
50–90 kV |
90–160 kV |
225 kV |
Laser |
RM 100 PL |
✓ |
|
|
|
|
|
|
RM 110 PL |
|
|
✓ |
✓ |
|
|
|
RM 200 PL |
✓ |
|
|
|
|
|
|
RM 210 PL |
|
|
✓ |
✓ |
✓ |
|
|
LaserTX |
|
|
|
|
|
|
✓ |
RM 200 CM |
✓ |
|
|
|
|
|
|
RM 210 CM |
|
|
✓ |
✓ |
✓ |
|
|
RM 215 HM |
|
|
|
|
✓ |
✓ |
|
RM 302 ES |
✓ |
|
|
|
|
|
|
RM 200 EG |
|
✓ |
|
|
|
|
|
RM 316 EG |
|
|
|
|
|
✓ |
|
SIPRO |
|
|
|
|
|
✓ |
|
RM 210 AF |
|
|
✓ |
|
|
|
|
RM 210 AS |
|
|
✓ |
|
|
|
|
RM 210 AH |
|
|
|
✓ |
✓ |