Simplicity, productivity and robustness for routine labs
Choose the Thermo Scientific iCAP RQ ICP-MS for a complete multi-element analysis solution for your high-throughput routine lab. With comprehensive interference removal for assured data accuracy and intuitive workflows to boost productivity, this single quadrupole (SQ) ICP-MS will expand your analytical capabilities.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Our user-inspired hardware and software maximizes ease-of-use and streamlines workflows to achieve 'right-first-time' analysis, essential to all busy laboratories.
Increase throughput, minimize sample re-runs and save analysts' valuable time and effort with intuitive features and broad compatibility with automated accessories.
- 'Get Ready' one-click set-up for easy, reproducible analysis
- Cone change in as little as 60 seconds
- Fast, easy operation with quick-connect sample introduction components
Comprehensive interference removal achieved with our innovative QCell flatapole design assures data accuracy. Using a single helium Kinetic Energy Discrimination (He KED) mode, all analytes can be measured at sub-ppt levels, even lithium and beryllium.
- Measure all analytes in a single helium Kinetic Energy Discrimination (He KED) mode
- In He KED mode, even lithium and beryllium can be measured at sub ppt levels
- Excellent interference removal with high ion transmission
- Reactive gas capability targets specific interference ions for challenging applications
Logical, problem-solving hardware ensures maximum up-time and minimum maintenance. Reliable plasma operation and high matrix tolerance let you run automated, unattended sampling, 24/7, with confidence. A choice of interface options, including our Robust Interface gives you the perfect choice for your sample type and analysis needs.
Interface | When | Why | Who |
---|---|---|---|
Robust | Long-term analysis of dirty, high matrix samples | Minimal drift and reduced user maintenance | Environmental and routine industrial applications |
High Matrix | Typical, everyday matrix types | Best balance for sensitivity and matrix tolerance | Food safety, pharmaceutical and drinking water analysis |
High Sensitivity | Advanced applications; e.g. laser, nanoparticles | Best signal to noise ration and lowest detection | Geoscience, advanced materials, research |
Intuitive workflows and intelligent features offered with Thermo Scientific Qtegra Intelligent Scientific Data Solution™ (ISDS) software, simplify method development, increase throughput and minimize reruns. Productivity is enhanced through a ranfe of automated sample handling options, all managed within a logical dashboard interface.
Improve performance, convenience and flexibility with inductively coupled plasma mass spectrometer consumables and accessories designed specifically for your instrument.