Search Thermo Fisher Scientific
Thermo Scientific Orbitrap Astral Mass Spectrometers are powered by unique architecture featuring a quadrupole mass analyzer and two high-resolution accurate mass (HRAM) analyzers: the Thermo Scientific Orbitrap analyzer and Thermo Scientific Astral analyzer. This powerful combination enables novel data acquisition strategies, including narrow-window data-independent acquisition (nDIA) and simultaneous quantitation and discovery (SQUAD) workflows, to deliver higher performance across many applications.
Accelerate discoveries in proteomics, metabolomics, lipidomics, structural biology, and biopharma applications with faster throughput, deeper coverage, higher sensitivity, and accurate and precise quantitation to achieve the next generation of breakthroughs.
For Research Use Only. Not for use in diagnostic procedures.