• Get fresh hepatocytes when you need them from the industry’s largest sourcing network
  • Standard animal species are available at least once a month, and fresh human cells are available weekly
  • Stringent release specifications ≥80% viability and ≥80% confluency (plated cells)
  • Ready to use - plated to your specifications or provided in suspension
  • GIBCO® Fresh Hepatocytes frequently asked questions
RapidAlert™:  Sign up to receive e-mail notifications when fresh hepatocytes become available.
Request more information
For information about fresh hepatocyte product, contact hepaticproducts@thermofisher.com
Availability
Animal (North America only):  Regularly scheduled isolations are detailed on our Animal Isolation Calendar.  Standard isolations are from male animals, unless otherwise requested.  Upon request, custom isolations can be scheduled.
Human:  The supply of fresh human hepatocytes is dependent upon availability of human liver tissue.  Register in our RapidAlert™ system for immediate notification when fresh human hepatocytes become available.
To place an order or make special shipping arrangements, please contact us at  hepaticproducts@invitrogen.com or (866) 952-3559
Primary Hepatocytes Isolated from Fresh Human & Animal Liver Tissue:
  • Human hepatocytes
  • Rat (Sprague-Dawley) hepatocytes
  • Mouse (CD-1) hepatocytes
  • Dog (Beagle) hepatocytes
  • Monkey (Cynomolgus) hepatocytes
Custom Species Available Upon Request
Formats
  • Available in suspension or in multi-well plates (6-, 12-, 24-, 48-, 96-well plates)
  • Available with a collagen type I substratum, with or without Geltrex® overlay
  • Special matrix configurations are available upon request
Plating density of Gibco® fresh human hepatocytes.
Plate format 6-well 12-well 24-well 48-well 96-well T75 flask
Million cells/mL ~0.85 ~0.67 ~0.60 ~0.50 ~0.31 ~1.33
Million cells/well 1.70 0.67 0.30 0.13 0.04 12.0
Fresh Hepatocyte Product Information
Product SKU
Human and Animal Fresh Suspension Hepatocytes
Human fresh hepatocytes, suspension HMFS01
Dog (beagle) fresh hepatocytes, suspension DGFS01
Monkey (cynomolgus) fresh hepatocytes, suspension MKFS01
Mouse (CD-1) fresh hepatocytes, suspension MSFS01
Rat (Sprague-Dawley) fresh hepatocytes, suspension RTFS01
Human and Animal Fresh Plated Hepatocytes
Human fresh hepatocytes, 6-well HMFN06
Human fresh hepatocytes, 6-well with overlay HMFY06
Human fresh hepatocytes, 12-well HMFN12
Human fresh hepatocytes, 12-well with overlay HMFY12
Human fresh hepatocytes, 24-well HMFN24
Human fresh hepatocytes, 24-well with overlay HMFY24
Human fresh hepatocytes, 48-well HMFN48
Human fresh hepatocytes, 48-well with overlay HMFY48
Human fresh hepatocytes, 96-well HMFN96
Human fresh hepatocytes, 96-well with overlay HMFY96
Dog (beagle) fresh hepatocytes, 6-well DGFN06
Dog (beagle) fresh hepatocytes, 6-well with overlay DGFY06
Dog (beagle) fresh hepatocytes, 12-well DGFN12
Dog (beagle) fresh hepatocytes, 12-well with overlay DGFY12
Dog (beagle) fresh hepatocytes, 24-well DGFN24
Dog (beagle) fresh hepatocytes, 24-well with overlay DGFY24
Dog (beagle) fresh hepatocytes, 48-well DGFN48
Dog (beagle) fresh hepatocytes, 48-well with overlay DGFY48
Dog (beagle) fresh hepatocytes, 96-well DGFN96
Dog (beagle) fresh hepatocytes, 96-well with overlay DGFY96
Monkey (cynomolgus) fresh hepatocytes, 6-well MKFN06
Monkey (cynomolgus) fresh hepatocytes, 6-well with overlay MKFY06
Monkey (cynomolgus) fresh hepatocytes, 12-well MKFN12
Monkey (cynomolgus) fresh hepatocytes, 12-well with overlay MKFY12
Monkey (cynomolgus) fresh hepatocytes, 24-well MKFN24
Monkey (cynomolgus) fresh hepatocytes, 24-well with overlay MKFY24
Monkey (cynomolgus) fresh hepatocytes, 48-well MKFN48
Monkey (cynomolgus) fresh hepatocytes, 48-well with overlay MKFY48
Monkey (cynomolgus) fresh hepatocytes, 96-well MKFN96
Monkey (cynomolgus) fresh hepatocytes, 96-well with overlay MKFY96
Mouse (CD-1) fresh hepatocytes, 6-well MSFN06
Mouse (CD-1) fresh hepatocytes, 6-well with overlay MSFY06
Mouse (CD-1) fresh hepatocytes, 12-well MSFN12
Mouse (CD-1) fresh hepatocytes, 12-well with overlay MSFY12
Mouse (CD-1) fresh hepatocytes, 24-well MSFN24
Mouse (CD-1) fresh hepatocytes, 24-well with overlay MSFY24
Mouse (CD-1) fresh hepatocytes, 48-well MSFN48
Mouse (CD-1) fresh hepatocytes, 48-well with overlay MSFY48
Mouse (CD-1) fresh hepatocytes, 96-well MSFN96
Mouse (CD-1) fresh hepatocytes, 96-well with overlay MSFY96
Rat (Sprague-Dawley) fresh hepatocytes, 6-well RTFN06
Rat (Sprague-Dawley) fresh hepatocytes, 6-well with overlay RTFY06
Rat (Sprague-Dawley) fresh hepatocytes, 12-well RTFN12
Rat (Sprague-Dawley) fresh hepatocytes, 12-well with overlay RTFY12
Rat (Sprague-Dawley) fresh hepatocytes, 24-well RTFN24
Rat (Sprague-Dawley) fresh hepatocytes, 24-well with overlay RTFY24
Rat (Sprague-Dawley) fresh hepatocytes, 48-well RTFN48
Rat (Sprague-Dawley) fresh hepatocytes, 48-well with overlay RTFY48
Rat (Sprague-Dawley) fresh hepatocytes, 96-well RTFN96
Rat (Sprague-Dawley) fresh hepatocytes, 96-well with overlay RTFY96