Accelerate your oligonucleotide synthesis and development with a wide variety of Thermo Fisher Scientific phosphoramidites and tools. Our expert product support team is available to guide your screening, development, manufacturing and beyond, ensuring you have the most efficient and reproducible path to commercialization. Take your oligonucleotide synthesis project from early research to preclinical candidate with a single supplier that understands your needs every step of the way.
Oligonucleotide synthesis
Oligonucleotide therapeutics are a new class of drugs comprised of short strings of synthetic nucleotides. Oligonucleotide synthesis is a chemical pathway sequentially incorporating phosphoramidites to grow specific sequences of modified and unmodified DNA or RNA. The resulting molecules can then be used in a variety of applications including forensic and diagnostic assays (PCR, qPCR, and other platforms), nucleic acid therapeutics (RNAi, antisense, aptamer, CRISPR), and basic research (Figure 1).
Figure 1. A schematic diagram outlines the phosphoramidite oligonucleotide solid phase synthesis method.
Reagents for oligonucleotide synthesis
Thermo Fisher Scientific offers a range of phosphoramidites and solid supports for oligonucleotide synthesis:
DNA Phosphoramidites
- TheraPure DNA phosphoramidites, the original brand of amidites designed specifically for therapeutic strategies
- Continued development focuses on demand for high-sensitivity assays and next-generation therapeutic molecules
RNA Phosphoramidites
- TheraPure RNA phosphoramidites are the gold standard for development and manufacturing of RNA-based therapeutic oligonucleotides
- Includes 2’-modified RNA phosphoramidites to help increase stability and improve resistance to nuclease activity in vivo
2’ O-methyl (OMe) RNA Phosphoramidites
- Traditional staple for oligo therapeutics for many years
- Support novel technologies such as CRISPR-Cas9 gRNA
- Key component of oligonucleotides designed for antisense, siRNA, aptamer, or gRNA-based applications
- Modifications offer:
- Nuclease resistance
- Fewer off-target effects
- Increased hybridization affinities
- Faster deprotection with modified version including PAC-A and iPrPAC-G
- Nuclease resistance
2’ O-methyoxy-ethyl (MOE) RNA Phosphoramidites
- Advanced generation of antisense oligonucleotides and siRNA designs
- Key component of antisense-based oligonucleotides
- Modifications offer:
- Nuclease resistance
- Fewer off-target effects
- Increased hybridization affinities
- Nuclease resistance
2’ Fluoro (Fl) RNA Phosphoramidites
- Modifications support stability and affinity for oligos used in therapeutic strategies
- Recently upgraded to meet standards of TheraPure products
- Modifications offer:
- Nuclease resistance
- Increased thermal stability of RNA duplexes
Linkers and Spacers
- 3′, 5′, and internal oligo linkers are modifications used for NHS ester conjugation
- Spacers increase the distance between functional groups, and provide conformational support
Structural Phosphoramidites
- Used for molecular diagnostics and research applications
- Modifications for design of primers, probes, or for structural studies
- Modifications includes 5m-dC, dU, dI, N6-Me-dA, N4-Ethyl-dC, 8-oxo-dG, and more
Customization for oligonucleotide synthesis
Custom chemistries (e.g., triphosphate, phosphoramidites, and custom organic), customized QC, and packaging options including filling and labeling are available.
- Dimethoxy (DMT) substitution with alternate protecting groups
- Base = A, C, G, T, or U protection modifications or base modifications
- Sugar 2’ position modifications and protection options
- Phosphoramidite, succinate, and support attachment and proprietary chemistries
Thermo Scientific TheraPure phosphoramidites
Originally designed specifically to meet the needs of oligonucleotide synthesis and development, TheraPure phosphoramidites are now important for therapeutics, molecular assay, and gene synthesis applications, which benefit from low levels of critical impurities. Our Thermo Scientific TheraPure phosphoramidites are designed for oligonucleotide manufacturers requiring highly defined impurity profiles and strictly controlled processes.
DNA Phosphoramidites
Click images to enlarge
dA(Bz) CEP | dC(Bz) CEP | dG(iBu) CEP | dT CEP | |
---|---|---|---|---|
Cat. No. | 27-2030 | 27-2032 | 27-2034 | 27-2036 |
Formula | C47H52N7O7P | C46H52N5O8P | C44H54N7O8P | C40H49N4O8P |
RNA Phosphoramidites
Click images to enlarge
2’-MOE RNA Phosphoramidites
Click images to enlarge
2′-MOE-A CEP | 2′-MOE-5mC CEP | 2′-MOE-G CEP | 2′-MOE-T CEP | |
---|---|---|---|---|
Cat. No. | 27-1019 | 27-1020 | 27-1022 | 27-1021 |
Formula | C50H58N7O9P | C50H60N5O10P | C47H60N7O10P | C43H55N4O10P |
2’-Fluoro RNA Phosphoramidites
Click images to enlarge
2′-Fluoro-Bz-A-CEP | 2′-Fluoro-iBu-G-CEP | TheraPure 2′-Fluoro-Ac-C-CEP | TheraPure 2′-Fluoro-U-CEP | TP 2'-Fluoro-Ac-G-CEP | |
---|---|---|---|---|---|
Cat. No. | 27-1601 | 27-1607 | 27-1604 | 27-1602 | SY5410* |
Formula | C47H51FN7O7P | C44H53FN7O8P | C41H49N5FO8P | C39H46FN4O8P | C42H49FN7O8P |
* For more specification details, contact us.
2’ O-methyl (OMe) RNA Phosphoramidites
Click images to enlarge
2′-OMe-Bz-A-CEP | 2′-OMe-U-CEP | 2′-OMe-iBu-G-CEP | 2′-OMe-Ac-C-CEP | |
---|---|---|---|---|
TheraPure phosphoramidites' Cat. No. | 27-2042 | 27-2044 | 27-2046 | 27-2043 |
Formula | C48H54N7O8P | C40H56N7O9P | C45H56N7O9P | C42H52N5O9P |
Enabling oligonucleotide synthesis and development
Manufacturing excellence and expertise
With over 4 decades of engineered nucleic acid manufacturing experience, phosphoramidites combine our expertise in protection chemistries, modifications, manufacturing scale-up, and innovative purification strategies. All catalog (and custom) phosphoramidites are manufactured in our state-of-the-art Milwaukee, WI manufacturing facility, with an ISO 9001:2015 certified quality management system.
Quality manufacturing and regulatory documentation
Processes are monitored from material procurement through final QC and product release, providing batch-to-batch reproducibility. Our rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide during synthesis and result in the accumulation of by-products. TheraPure phosphoramidites from Thermo Fisher Scientific are designed for pharmaceutical manufacturers looking for core building blocks to use in research and manufacturing. These reagents are crafted in facilities with thoroughly documented quality systems and audited by accredited registrars.
Interview videos
Watch our scientists discuss a variety of topics covering Thermo Fisher Scientific's Milwaukee site history, customer partnership, manufacturing process, quality improvement, mitigation of supply chain issues, and response to global needs.
Resources
Brochure
Flyer
Guide
Technical note
Poster
eBook
For Research Use Only. Not for use in diagnostic procedures.