Select an article below to learn how to create and share reports of your sample data.
Generate reports from templates to quickly save or share your data as Microsoft™ Word™, PowerPoint™, or Excel™ files, or export your spectra, workflows, or settings.
To create a report
- From the spectral view, open the menu and select Create Report.
- In the Output As dialog box, enter a title for your report, select a directory to save the report to, choose a format, and finally select a template. The preview updates to match your format and template selection.
If you select Save as your format, your report data is saved to your database.
- Touch Create.
To create a report
- Select File > Create Report.
- In the Output As dialog box, enter a title for your report and select a format and template option.The preview updates to match your selections.
If you select Save as your format, your report data is saved to the database. - Click Create.
To create a report
- From the spectral view, open the menu and select Create Report.
- In the Output As dialog box, enter a title for your report, select a directory to save the report to, choose a format, and finally select a template. The preview updates to match your format and template selection.
If you select Save as your format, your report data is saved to your database.
- Touch Create.
To create a report
- Select File > Create Report.
- In the Output As dialog box, enter a title for your report and select a format and template option.The preview updates to match your selections.
If you select Save as your format, your report data is saved to the database. - Click Create.
Export Settings, Spectra, or Workflows
Use OMNIC Paradigm software for desktop to export settings and workflows. Export spectra in either desktop or touchscreen mode.
To export settings
- Select the settings you wish to export from the Settings list.
- Select Acquire Data > Export Settings.
- Name the settings file and click Save.
Settings are saved with a .expx file extension. These settings files store your measurement settings but not Search, QCheck, or Quantification settings. Settings files with a .expx file extension are not compatible with previous versions of OMNIC software.
To export workflows
- In the Workflows pane, right-click the workflow you wish to export and select Export from the menu.
- Name your workflow file and click Save.
Workflow files are saved with a .rwfl file extension.
To export a spectrum
- From the home screen, select the Measurements tab.
- Open the more options menu [
] and select Export.
- Touch Save. Spectra are saved with a .spa file extension.
To export a spectrum
- In the Measurements pane, right-click the spectrum you wish to export and select Export.
- Name your spectrum file and click Save. Spectra are saved with a .spa file extension.
To export a spectrum
- From the home screen, select the Measurements tab.
- Open the more options menu [
] and select Export.
- Touch Save. Spectra are saved with a .spa file extension.
To export a spectrum
- In the Measurements pane, right-click the spectrum you wish to export and select Export.
- Name your spectrum file and click Save. Spectra are saved with a .spa file extension.