Nicolet Summit FTIR Spectrometer
Select an article below or watch a video to learn how to install and use the Thermo Scientific OMNIC Paradigm Software for FTIR spectroscopy.
Quick links
OMNIC Paradigm Software for desktop and touchscreen is an advanced software package for FTIR spectroscopy, designed to simplify how you acquire, process, and interpret data and to help you work remotely and collaborate with colleagues around the globe.
Manage new and recent work from the dashboard
welcome-omnic-paradigm-software-image-710x575
With the new dashboard in OMNIC Paradigm Software, you can set and save your measurement settings, measure a new sample, resume your recent work, or run an automated workflow all from the same, convenient screen.
In Paradigm for touchscreen, the home screen provides many of these same tools and features in an even more streamlined, intuitive interface so operators can focus on their work and on the tools they need.
Process, analyze, and explore spectra in the spectral view
welcome-omnic-paradigm-software-image-710x443
Use the spectral view to process, analyze, or explore your data in depth. Here you can process your data, find and label peaks, and optimize the view so you can more easily compare or analyze spectra.
The results pane allows you to quickly add or remove a spectrum from the main view, explore details of each spectra, or review the change history of your selected spectrum.
Automate repetitive tasks with workflows
welcome-omnic-paradigm-software-image-710x444
Automate your tasks with a workflow to ensure that measurements are collected and processed in exactly the same way each time. Edit or create a workflow by simply dragging and dropping tiles in the workflow editor, and then run the workflow at the click of a button.
Operators using Paradigm for touchscreen can import and run one of our factory installed performance tests or a custom workflow right from the home screen.
Share your work remotely
welcome-omnic-paradigm-software-image-710x384
Upload your data to the cloud and use OMNIC Anywhere to view, analyze or share your data using any connected PC, Apple computer, Android or iOS device.
With a Thermo Scientific Nicolet Summit Spectrometer and a free Connect account, measure samples in the classroom or lab, upload the data to your Connect account, and view, explore, or share the data on another device in your dorm or workspace.
The latest version of OMNIC Paradigm software features powerful new ways to process data, updated workflow tiles, and the ability to customize your reports with your organization's logo.
For more details about what's new in version 1.9 HF 1 of OMNIC Paradigm software, download the release notes
Reprocess spectra
Use Reprocess in the Process menu to transform the original interferogram data for your spectrum with new settings. With Reprocess, you can transform your original data to better match library or other reference spectra, giving you a more accurate comparison.
omnic-paradigm-reprocess
Blank regions of spectra
With Blank Regions in the Process menu, you can clear unwanted data from your spectrum. Blanking a region is useful when your spectrum contains unwanted noise that would otherwise interfere with your analysis. Use Blank Regions to clear, or delete, a region of the spectrum. Blank Regions is also available in the Processing tile in Workflows.
omnic-paradigm-blank-regions
Find the average and standard deviation of spectra
You can now calculate average and standard deviations of spectra using the Statistical Spectra tool in the Process menu. With Statistical Spectra, you can generate new spectra that represent the average or standard deviation of 2 or more spectra. You can also use Statistical Spectra in the Processing tile in Workflows.
Add custom logos to reports and template reports
Go to Configure > Options to use your own company logo in reports and template reports. Your logo replaces the default Thermo Scientific logo on any custom reports you create and any template reports created in a workflow.
Improvements to workflows
With the Peak Analysis tile and Process tile in Workflows, you can now use Find Peaks,  Blank Regions, and calculate statistical spectra in your workflows.
OMNIC Paradigm Software comes preinstalled on the Nicolet Summit Spectrometer and is ready to use with the attached touchscreen monitor or with an external monitor connected by Mini DisplayPort.
Install OMNIC Paradigm software on your desktop, laptop, or other device to connect to Summit spectrometers or other instruments, such as Nicolet iS5, over an Ethernet, WiFi, or USB connection. Connect to an instrument to measure a sample and disconnect to allow other operators to use the instrument while you analyze your data or develop workflows. 
This guide walks you through installing OMNIC Paradigm software. 
Before you begin
Your system should meet the recommended system requirements outlined below to run and use OMNIC Paradigm Software.
Recommended system requirements
Operating systemWindows 10 (64-bit)
ProcessorIntel Core i5 3.3GHz
RAM8 GB
Hard disk
2 GB of available hard-disk space for installation; Additional hard-disk space recommended for storing files.
If you are hosting your database on this machine, we recommend 500 GB of hard-disk space.
Install OMNIC Paradigm software
Installing OMNIC Paradigm takes only a few minutes.
To install OMNIC Paradigm software
  1. Insert the provided installation media (either DVD or flash drive).
    1. Install OMNIC Paradigm Software
  2. Run the installer.
    1. When the download completes, run the Start.exe file.
    2. Follow the on-screen prompts to install OMNIC Paradigm software.
Set up and connect OMNIC Paradigm workstation
By default, data is saved to the database on your local device. If you connect to a Nicolet Summit Spectrometer, you can continue using your local database, or you can connect to the database on the instrument or another database located on your network. 
To connect to a default database
  1. Open OMNIC Paradigm workstation software.
  2. Select Configure > Database.
  3. In the Database Engine list, select Thermo Scientific Built-in Database.
  4. Enter the server address or URL for your database. The server address or URL is the name of the instrument that you want to connect to on your network.
OMINIC Paradigm Software features three interfaces to suit your device or task and pairs with other Thermo Scientific software, allowing you to develop and run quantification methods or protect and audit your data.
OMNIC Paradigm Software interfaces
Switch anytime between OMNIC Paradigm Software’s touchscreen and desktop interfaces to best suit your device and goals.
Touchscreen interface
Figure 1. The touchscreen interface features a simplified interface for streamlined analysis.
welcome-omnic-paradigm-software-image3-710x427
The touchscreen interface is optimized for small screens, such as the Nicolet Summit Spectrometer’s optional touchscreen device.
Use the touchscreen interface for measuring and analyzing samples or running workflows in a streamlined, simplified interface.
Desktop interface
Figure 2. The desktop interface includes a wider range of features and tools for more complex measurements and analyses.
welcome-omnic-paradigm-software-image3-710x575
Use the desktop interface when working from a larger screen and when you need additional features not available in the touchscreen interface, such as the workflow editor and select spectral processing and analysis tools.
Connect your Summit Spectrometer to an external monitor and a USB mouse and keyboard for the best experience using the desktop interface.
Operator interface
The Operator interface is used for running workflows loaded from packages and for managing packages. After opening a package, operators are presented with a simplified interface designed to run the package workflows.
paradigm-operator-interface
For details on using the Operator interface and developing custom solutions, see "Custom Solutions".
Switching between interfaces
Although desktop and touchscreen software are optimized for different tasks and devices, you can switch between interfaces at any time.
To switch between the touchscreen and desktop interfaces
  • To switch from desktop to touchscreen, select View / Display > Switch to Touchscreen.
    The software will close briefly and re-open with the touchscreen interface.
  • To switch from touchscreen to desktop, from the home screen, select Switch to Desktop from the menu.
    The software will close briefly and re-open with the desktop interface.
Pairing OMNIC Paradigm with other Thermo Scientific software
OMNIC Paradigm software is compatible with other Thermo Scientific software for added flexibility and for quantitative analysis and data security.
OMNIC Paradigm Workstation software
Figure 3. Workstation software lets you analyze spectra or develop workflows while you’re away from a spectrometer
welcome-omnic-paradigm-software-image3-710x442
When working from a separate computer and not connected to an instrument, use Workstation software to analyze previously collected data, create workflows for later use, or create and share reports.
Quantitative analysis software
Pair OMNIC Paradigm software with Thermo Scientific TQ Analyst Software version 9.8 or later to develop and run quantitative analysis methods that you can use in OMNIC Paradigm Software.
  • TQ Analyst EZ Edition
  • TQ Analyst Professional Edition
See “TQ Analyst Pro Edition Software” on thermofisher.com for details.
Data security software
Use Thermo Scientific OMNIC Security Suite Software version 2.2 or later to ensure your data’s security and integrity. The Security Suite Software ensures 21CFR Part 11 compliance and includes the following:
  • Security Administration for controlling system access, handling security policies, and setting digital signatures for responsibility and data integrity
  • Audit Manager for creating and managing complete audit trails
  • Thermo Software IQ tool to verify your security installation
Configure your database
When you first install OMNIC Paradigm software, the default database is set up on the local device and ready to use. You can continue to use the local database or connect to a remote database in another location on your network. Use a remote database to easily share measurements, settings, and other data across devices.
When using OMNIC Paradigm Workstation software, if you connect to a Nicolet Summit spectrometer, data will be stored in the database configured in the workstation software, not the database configured on the spectrometer.
Connect to a remote database
Connect to a remote database to easily share measurements, settings, and other data across devices. Change your database configuration at any time to suite your work.
To connect to a remote database
  1. Using the desktop interface of OMNIC Paradigm software, go to Configure and select Database.
    Screenshot of the database configuration window
  2. Select the database type from the Database list. The default selection is Thermo Scientific Built-in. To connect to a custom database, enter the database type, version, name, and port. You may also need to enter a username and password.
  3. Enter the IP address or URL for the database server.
  4. Click Test Connection to check if the software can connect to the server.
  5. If the test reports a valid connection, click OK to connect to the remote database. The software restarts during connection.
Check the Measurements group on the dashboard to confirm that you are seeing data from the remote database, or return to the Configure > Database menu to view your current connection.
Back up and restore a database
Back up your database periodically to ensure that you can recover your data in the event of hard drive failure. With the database maintenance tools in OMNIC Paradigm software, you can easily backup and restore your data directly from the software.
Back up your database
Back up your database on a separate device in case of hard drive failure. For example, back up the data on a separate computer or external hard drive or in a remote network location.
To back up your database
  1. Using the desktop interface of OMNIC Paradigm software, go to Configure > Database Maintenance and select Back Up.
    Screenshot of the database maintenance back up window
  2. Select a backup location to save the backup database.
  3. To schedule backups, set a date and time for the first backup.
    1. Ensure that Repeat is selected and choose a repeat option from the list.
    2. Click Save.
  4. To back up the database manually without a schedule, click Back Up Now
    To set a new backup location before backing up, first change the location and save your changes, then reopen the Database Maintenance - Back Up window and click Back Up Now.
  5. Click Close to return to the dashboard.
Restore a database from a backup
Use the Restore tool in OMNIC Paradigm software to restore your database from a previously saved backup.
Notice This action will permanently remove all data currently in your database and will replace it with data from the selected backup.
To restore your database
  1. Using the desktop interface of OMNIC Paradigm software, go to Configure > Database Maintenance and select Restore.
    Screenshot of the database maintenance restore window
  2. Click Browse and select the location of the backup database.
    If you are using OMNIC Paradigm software on a Summit spectrometer and you are restoring from a remote backup that is shared with other spectrometers, you can restore data from any individual spectrometer that is using the backup.
    To back up data from only a single spectrometer, select a spectrometer from the list of hostnames.
  3. Click Confirm.
Understanding the core concepts of using OMNIC Paradigm Software will help you more efficiently and effectively collect, analyze, and report spectral data.
You may not always carry out every step of the following process, and you can customize OMNIC Paradigm Software to meet your specific needs, but in general, you will likely follow a common process:
welcome-omnic-paradigm-software-image1-690x101
Plan Your Project
welcome-omnic-paradigm-software-image2-690x101
Before getting started with the software, it’s important to consider your project goals. Planning in advance will help you decide what type of processing and analysis is appropriate for your work, what measurement settings will yield the best data, and how you can most efficiently carry out your procedures.
Before acquiring data, you should be able to answer the following questions:
  • What are my goals for the project?
    For example, do you want to identify an unknown sample or verify that your sample is meeting your standards. With OMNIC Paradigm Software, you can run different processes and analysis steps depending on how you will use the data. Common goals include the following:
    • Identify a pure sample
    • Identify the components of a mixture
    • Verify that a sample meets your specifications
    • Quantify the components of a sample
  • How will I measure my sample?
    OMNIC Paradigm software supports a variety of spectrometer sampling accessories and measurement techniques.
    See “FTIR Sample Handling Techniques” on FTIR Spectroscopy Academy for a full discussion of choosing the best collection method for your specific work.
  • Am I planning only a few measurements or will this be a routine, repeated procedure?
    If you would like to automate a routine procedure, you can create a workflow. With a workflow, you specify settings for data collection, provide instructions to operators, and define data processing, reporting, and archiving in advance. You or other operators can then use the workflow to carry out procedures with the knowledge that exactly the same steps will be taken each time.
    For a step-by-step guide on creating a new workflow, see “About Workflows”.
Acquire data
welcome-omnic-paradigm-software-image3-690x101
Once you have a plan for collecting and working with your data, you’re ready to start measuring samples. Generally, to measure a sample and collect data, you will follow these steps:
  1. Prepare the spectrometer, sampling accessory, and sample to be measured.
    Usually, you should make sure your spectrometer, sampling accessory, and sample are ready before you begin. This can mean ensuring the spectrometer is warmed up and functioning well, that the sampling accessory is installed, and that the sample is prepared.
    The specific steps for setting up the spectrometer and accessory will vary depending on the instrument and on your sample. See the user guides and tutorials for your instrument for specifics on preparing your spectrometer. Or, see FTIR Spectroscopy Academy for more on handling and measuring samples.
    Note that you must measure the background without the sample present. The best time to collect a background spectrum will vary by the type of sample you are measuring.
  2. Select or edit your measurement settings.
    When the instrument is ready, you can adjust settings both for how the software will collect and process the data and for setting up the spectrometer. Select your saved settings from a menu on the dashboard or edit settings individually for a single measurement.
    See “Measurement Settings,” for a discussion of how to edit and save measurement settings.
  3. Measure the background.
    When you measure a sample, the spectrometer records data from the sample as well as from the environment in the spectrometer. To ensure that your sample spectrum represents only your sample, without any data from the background environment, you must measure the background. The background spectrum is then compared to (or ratioed against) the sample spectrum and any data from the background is accounted for, leaving you with data from only your sample.
    See “Measure the Background,” for a full discussion of when and how to collect a background spectrum.
  4. Measure the sample.
    Finally, you can measure the sample. During the measurement, you can preview the spectrum and refine your measurement settings before proceeding. If you indicated an analysis type in the settings, the data will automatically be processed after collection.
    Once the data are collected, you will be able to view and explore the spectrum in the spectral view.
Process data
welcome-omnic-paradigm-software-image4-690x101
After you measure your sample, it’s often helpful to process your data so that it is easier to analyze or compare to other spectra. Common spectral issues that OMNIC Paradigm software can help you correct include the following:
  • A sloping, curving, shifted or otherwise undesirable baseline
  • Peaks obscured by noise in the data
  • Variation in depth of penetration from an ATR measurement
  • Totally absorbing peaks from too thick of a sample
  • Issues caused by CO2 or H2O in the atmosphere
Analyze the sample
welcome-omnic-paradigm-software-image5-690x101
OMNIC Paradigm software has tools designed to help you carry out common analyses and to explore spectral data in-depth. Common types of analysis include the following:
  • Identify an unknown sample or mixture
    To identify an unknown sample or the components of a mixture, compare the spectrum of your sample to reference spectra that are stored in spectral libraries. Spectral libraries are collections of spectra and usually contain additional information on the stored compounds. Libraries can be created or they can be purchased. When you run a correlation or multi-component search, the software searches your stored libraries and finds spectra that are similar to your sample spectrum and indicates how closely the spectra match.
    For more on spectral libraries, see “Manage Libraries.”
    For more on identifying an unknown sample, see “Identify an Unknown with ATR.”
  • Verify the composition of a sample
    You can quickly verify that a sample meets your specifications by using QCheck. With QCheck, you can compare two selected spectra or compare one or more spectra with a reference spectrum or spectral group. QCheck results show the degree of similarity between the spectra as a match value from 0.0 (no similarity) to 1.0 (the spectra are identical).
  • Find the concentrations of components in a sample
    Quantitative analysis methods that you have created in TQ Analyst Software are available for use in OMNIC Paradigm Software. You can select these quantification methods and run the quantification analysis from the OMNIC Paradigm dashboard.
  • Perform custom, in-depth analysis
    OMNIC Paradigm Software features many tools to help you to explore the spectra further. For example, the Find Peaks tool lets you quickly identify and label peaks, or you can subtract one spectrum from another or perform other operations using Spectral Math.
Generate reports and save data
welcome-omnic-paradigm-software-image6-690x101
Once you have collected, processed, or analyzed your data, you’ll likely want to store or share it. With OMNIC Paradigm software, you can customize how your data is stored or exported.
  • Exporting reports
    When it’s time to share your data, you can generate reports automatically using templates. Templates can include results from library searches, QCheck, quantification methods, and more, or they can be used to send data directly to Microsoft Word, Excel, or PowerPoint.
  • Storing data
    By default, the software is configured to store data in a MariaDB Server database, but you can also configure the software for other databases, such as Oracle Database or Microsoft SQL Server.
    Additionally, settings, spectra, and workflows can be exported as files that you can backup, store, or share.
    For a discussion of data security options in OMNIC Paradigm Software, see "21 CFR Part 11 Compliance."
Next steps
You may not always follow every step of the process described in this guide, but in general, you will be able to achieve better results by understanding the overall process and capabilities of using OMNIC Paradigm Software.
Next, get started by measuring your first samples, reviewing setup of your spectrometer, or creating a workflow to automate routine procedures.
There are three ways to connect your laptop or other device to your spectrometer. The connection you choose depends on the instrument.
  • USB
  • Network connection
    • local area network
    • wireless network
  • Direct Ethernet connection
See the user guides and help content for your instrument for more on setup, installation, and connecting to other devices.
Connect to an instrument using a USB connection 
Some instruments can be connected using an USB connection. For example, the Nicolet iS5 and Nicolet Summit LITE spectrometers use this type of connection.
To connect to an instrument using a USB connection
  1. Using a USB cable, connect the spectrometer to your computer running OMNIC Paradigm software.
  2. In OMNIC Paradigm software, go to Configure and select Connectivity.
  3. Select the spectrometer from the list of instruments and click Connect.
Connect to an instrument using an Ethernet or wireless network connection
You can connect to the Nicolet Summit, Summit PRO , or Summit OA spectrometer using an ethernet or wireless network connection. Before you connect to an instrument, make sure you understand database usage and user authentication.
Database usage
When you connect to an instrument, you will always use the database configured by the client system.
For example, if you are using OMNIC Paradigm software on a laptop and you connect to a Summit spectrometer, you will continue to use the database configured by the software on the laptop. When you disconnect from the spectrometer, the software on the laptop will still show the data that was collected while connected to the spectrometer. If you then work directly on the Summit spectrometer, you will not see any of the data that was collected with the laptop.
When using a shared remote database, the data that is visible depends on which system you are using. For example, if several Summit spectrometers share a remote database with OMNIC Paradigm software that is being run on a laptop, you can use the laptop to view data collected by any of the instruments. However, any individual spectrometer will show only data collected on that machine and not from any other spectrometer.
User authentication
When you connect to a Summit spectrometer, the spectrometer needs to authenticate your device to ensure the connection is safe and trusted. Many organizations manage authentication between devices by using a Windows domain. A domain is a group of computers that share user account information and security settings. With a domain, an administrator can manage many devices, accounts, and policies from a single server called a domain controller. This often allows you to log into many devices on the domain using the same username and password.
When you connect to a spectrometer from your laptop, the spectrometer and your laptop both communicate with the domain controller to make sure that the connection is safe and trustworthy. If either device is not on the domain, you will not be able to make the connection. 
If both devices are managed by the same domain, use a local or wireless network connection to connect to the spectrometer.
To connect to a Nicolet Summit spectrometer on a local area network or a wireless network 
If your computer and spectrometer are part of a shared domain (recommended), connect using WiFi or Ethernet. Before you begin, make sure the Summit spectrometer and the computer are on the same Ethernet or WiFi network.
  1. In OMNIC Paradigm software, go to Configure and select Connectivity.
  2. Select a spectrometer and click Connect.
If your spectrometer does not appear on the list of available instruments, you can also enter the IP address or hostname of the instrument and then click Connect.
If the spectrometer is already in use when you attempt to connect, the connection may be denied. The request can be denied for two reasons:
  • The instrument is busy measuring a sample or processing data and cannot be interrupted
  • Another user is operating the instrument and chose not to relinquish control
OMNIC-Paradigm-touchscreen-connections-request-710
If the current user does not respond within 30 seconds of the request, the connection is allowed automatically.
If the request is denied automatically or by the current user, you can return to Workstation mode, connect to another instrument, or try to connect again.
Connect directly to the instrument with an Ethernet cable
You can also connect to the spectrometer directly using an Ethernet cable. Using a direct connection requires you to edit the local security policies on the spectrometer. Because the instrument cannot communicate with a domain controller for user authentication when you are using this configuration, you must modify security policies to allow communication between the instrument and your computer. 
Note: Modifying security policies could pose a security risk to your organization. Follow your organization’s security policies.
To use a direct connection
  1. Modify security policies on the instrument
    1. On the Nicolet Summit spectrometer, open the Local Security Policy application.
    2. Navigate to Security Settings > Local Policies > Security Options. 
    3. Set Network access: Let Everyone permissions apply to anonymous users to Enabled. 
  2. Use an Ethernet cable to connect the computer directly to the spectrometer.
  3. In OMNIC Paradigm software, if the software does not recognize the instrument automatically, go to Configure > Connectivity and connect to the spectrometer. 
Preventing Connections from Unsecured Devices
If your spectrometer is using Thermo Scientific Security Suite software to protect and secure your data, you may want to prevent connections from unsecured devices. For example, with the default
settings, if a laptop that is not managed by Security Administration software and Audit Manager software connects to a secure spectrometer, any action taken on the laptop would not be managed or
logged.
To prevent connections from unsecured devices, you must edit your firewall settings on the secure spectrometer. Disable all Inbound rules that belong to the group RabbitMQ or to OMNIC Paradigm.
See "Firewall Settings" for additional details on these rules. 
To disable inbound rules in Windows Defender Firewall
  1. Open Windows Defender Firewall with Advanced Security.
  2. In the left pane, navigate to Inbound Rules.
  3. In the primary pane, under Inbound Rules, select a rule to disable.
  4. In the Actions pane, click Disable Rule.
The Enabled column for the rule you have selected changes to No.
To re-enable the rule, click Enable Rule in the Actions column.
Troubleshooting
Instrument not appearing in available connections
If your spectrometer does not appear automatically in the list of available instruments when you go to Configure > Connectivity, check your network connections.
The spectrometer and the computer making the connection must be on the same network for the instrument name to appear in the list. For example, if your organization has several wireless networks, make sure that the spectrometer and computer are on the same network.
If the spectrometer’s name doesn’t appear in list of instruments, you can attempt to connect by entering the spectrometer’s IP address.
Advanced troubleshooting
If you cannot connect to a spectrometer, you may need to check your firewall settings. Only the IT or network administrator at your organization should edit firewall settings.
Firewall settings
When OMNIC Paradigm software is installed, your firewall settings are updated to allow a connection between a spectrometer and OMNIC Paradigm Workstation software. If you experience problems with your connection, check your firewall settings on both the spectrometer and the connecting computer to confirm that the following ports allow traffic.
The table below reflects the inbound rules as set in Windows Defender Firewall. The available fields may be different depending on the application your organization uses to manage network firewall settings.
Table 1. Required inbound rules
NameDescription and ports used
Password management (NB-Name-In)
Description: Inbound rule for Password Management to allow NetBIOS Name Resolution.
Group: Thermo Scientific Password Management
Program: System
Ports: UDP 137
Action: Allow
Profile: Domain, Private, Public
Password management (NB-Session-In)
Description: Inbound rule for Password Management to allow NetBIOS Session Service connections.
Group: Thermo Scientific Password Management
Program: System
Ports: TCP 139
Action: Allow
Profile: Domain, Private, Public
Password management (SMB-In)
Description: Inbound rule for Password Management to allow Server Message Block transmission and reception via Named Pipes.
Group: Thermo Scientific Password Management
Program: System
Ports: TCP 445
Action: Allow
Profile: Domain, Private, Public
Thermo Scientific instrument discovery
Description: Allows clients to discover this instrument
Group: OMNIC Paradigm
Program: C:Program Files\Thermo Scientific\IR Instrument Engine\Instrument.IR.Engine.Service.exe
Ports: UDP 50123
Action: Allow
Profile: Domain, Private, Public
Thermo Scientific Web API Server
Description: Allows clients to access a web API
Group: OMNIC Paradigm
Program: System
Ports: TCP 9004
Action: Allow
Profile: Domain, Private, Public
RabbitMQ - peer discovery (EPMD)
Group: RabbitMQ
Program: C:\Program Files\erl9.3\erts-9.3\bin\epmd.exe
Ports: TCP 4369
Action: Allow
Profile: Domain, Private, Public
RabbitMQ - client connection (erl)
Group: RabbitMQ
Program: C:\Program Files\erl9.3\erts-9.3\bin\erl.exe
Ports: TCP 5671, TCP 5672
Action: Allow
Profile: Domain, Private, Public
RabbitMQ - management console (erl)
Group: RabbitMQ
Programs:
•      C:\Program Files\erl9.3\erts-9.3\bin\erl.exe
•      C:\Program Files\erl9.3\bin\erl.exe
Ports: TCP 15672
Action: Allow
Profile: Domain, Private, Public
Thermo Scientific history WebAPI
Group: OMNIC Paradigm
Program: System
Ports: TCP 9002
Action: Allow
Profile: Domain, Private, Public
Thermo Scientific workflow engine
Group: Thermo Scientific Workflow Engine
Program: System
Ports: TCP 9090
Action: Allow
Profile: Domain, Private, Public
Local security policies
When connecting devices that do not share a common directory service (such as Active Directory or LDAP), you must edit the following local security policy on the spectrometer.
To edit the local security policy, you will need a Windows account with administrator privileges. To edit the policy, in the Windows task bar, type Local Security Policy and open the Local Security Policy application. Edit the following setting:
Security Settings > Local Policies > Security Options:
EnableNetwork access: Let Everyone permissions apply to anonymous users.
Connect your instrument to your Thermo Fisher Connect account to view and manage your instruments and data remotely.
For more information on Thermo Fisher Connect and managing your connected instruments, see the Help guide in the Thermo Fisher Connect application.
Before you begin
To connect your instrument to Thermo Fisher Connect, you need a Thermo Fisher Connect account. To create an account, visit https://www.thermofisher.com/us/en/home/digital-science/thermo-fisher-connect.html.
To connect your instrument to your Thermo Fisher Connect account
  1. Using the Desktop interface of OMNIC Paradigm software, from the Dashboard, go to Acquire Data > Diagnostics > Instrument Health.
  2. Optional: Opt-in to sharing instrument health data.
  3. Click Link Instrument.
  4. To complete the connection, scan the QR code using the Thermo Fisher Instrument Connect app on your mobile device or use the 6-digit code.
OMNIC Paradigm connect to Instrument Health
To use the 6-digit code
  1. Click Copy to copy the code to your clipboard.
  2. Go to Thermo Fisher Connect in your web browser.
  3. On the dashboard of the Thermo Fisher Connect web application, under My Instruments, paste or enter the 6-digit code and click Link.
Thermo Fisher Connect
The instrument is now displayed in the InstrumentConnect section of Thermo Fisher Connect. 
To scan the QR code
  1. Open the Instrument Connect app on your mobile device.
  2. Touch the + icon in the upper right corner of the screen to add an instrument.
  3. Select QR Code.
  4. Use your device to scan the QR code.
The instrument is linked and displayed in the mobile app and online.