When accuracy and performance count
Go from samples to answers quickly and easily. Thermo Scientific UV-Vis and Vis instruments are designed to streamline measurements with consistent, high-quality results, time after time. From classroom teaching, to routine measurements, to discovery of the next breakthrough material, our line of spectrophotometers are designed to fit into today's modern laboratory.
Featured UV-Vis and Vis categories
Support your teaching goals. The Thermo Scientific SPECTRONIC 200 spectrophotometer is the standard for routine measurements in instructional and analytical laboratories. Designed for simplicity and speed, the SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer that provides quick, easy measurements and years of trouble-free performance.
Built for real world laboratories and classrooms where rugged, easy-to-use instrumentation is a must, Thermo Scientific GENESYS spectrophotometers give you a range of solutions for routine analysis and specialized measurements.
Transform your UV-Visible experience with the Thermo Scientific Evolution Spectrophotometers. Simplified workflows, cutting-edge technology and an array of accessories consistently deliver the high-quality results you depend on.
Measure life science materials quickly and accurately using pre-programmed routines. A simplified user interface and a high-resolution, color touchscreen enables you to apply automation for high-throughput measurements with room-light resistance, allowing lid-open operation for improved speed and convenience.
Thermo Scientific NanoDrop UV-Vis spectrophotometers feature a patented sample-retention system that enables direct measurements of 1 μL samples without dilutions, and pre-programmed methods designed specifically for life scientists. Thermo Scientific NanoDrop One microvolume spectrophotometers with Thermo Scientific Acclaro Sample Intelligence technology help you understand the quality of your sample before you use it in downstream applications, bringing you one step closer to success.
Quickly and reliably make endpoint, kinetic, or spectral UV-Vis measurements in multiple formats: Cuvettes, 96- and 384-well plates, and µDrop plate with 16 samples as small as 2 µL quickly in one run. With intuitive touchscreen interface with pre-programmed routines, Thermo Fisher Connect cloud-based tools, additional ready-to-use protocols in the SkanIt Cloud Library, quick export to Microsoft™ Excel™, and no limit to the number of SkanIt software licenses for fast operation and analysis, the Thermo Scientific Multiskan Sky Microplate Spectrophotometer is poised to simplify your UV-Vis workflow.
With the ability to measure all the same sample formats and driven by the same intuitive SkanIt software mentioned above for the Multiskan Sky spectrophotometer, this modular, upgradeable multimode reader can meet your UV-Vis and fluorescence measurement needs now and future-proof your lab by allowing you to add capabilities later. Take advantage of the flexibility of having no limit to the number of SkanIt software licenses, utilize even more pre-built protocols in the SkanIt Cloud Library, and quickly analyze within SkanIt software or export to Microsoftâ„¢ Excelâ„¢ for further analysis.
Right color? Right taste?
Be sure with UV-Vis
Ensure batch-to-batch consistency with BeerCraft Software and GENESYS 50 or 150 UV-Vis Spectrophotometers. Tests are preprogrammed for fast, easy quality analysis.