aquaculture-showcase
Solutions designed for simple and complex mosaic genomes
Aquaculture is one of the fastest growing food industries in the world, yet it is estimated that only 10% of aquaculture production is based on genetically improved stocks. Pedigree in aquaculture species is often costly and difficult to obtain, making development of optimized selective breeding programs challenging. Thermo Fisher Scientific offers genotyping solutions that can be designed for simple and complex mosaic genomes found in economically important aquaculture species such as shrimp, salmon and shellfishes.
Quick links
AgriSeq genotyping-by-sequencing (GBS)
The Applied Biosystems AgriSeq HTS Library Kit is specifically designed for high-throughput preparation of amplicon libraries for targeted genotyping-by-sequencing (GBS) applications in agrigenomics.
AgriSeq HTS Library Kits to be used with custom AgriSeq aquaculture SNP panels
Axiom microarrays
Our array-based genotyping solutions are ideal for the identification, validation and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays or customize an array specifically for your study. Custom aquaculture microarrays can be designed using Axiom myDesign Genotyping Arrays.
Product
No. of markers
Array format
Cat. No.
Price
Axiom Salmon Genotyping Array (Salmo salar)
430,743
96-array
550540
Contact a sales representative ›
Axiom Catfish Genotyping Array
693,567
96-array
550704
Contact a sales representative ›
Axiom Coho Salmon Genotyping Array
200,000
96-array
550862
Contact a sales representative ›
Axiom Oyster Genotyping Array
55,575
96-array
550796
Contact a sales representative ›
Axiom Trout Genotyping Array
57,501
96-array
550468
Contact a sales representative ›
Axiom Trout Genotyping Array
57,501
384-array
550571
Contact a sales representative ›
Custom TaqMan SNP Genotyping Assays
These assays provide a highly flexible technology for detection of polymorphisms within any genome. Create your own assays by submitting target sequences to our secure assay design pipeline using the Applied Biosystems Custom TaqMan Assay Design Tool. This pipeline has successfully generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. TaqMan Assays can be run on any of our real-time PCR instruments or used as an endpoint assay on other third-party systems, including the Array Tape platform.
Recommended reagents