companion animals
Innovative technologies for companion animal genomic testing
Choose from a variety of innovative technologies for companion animal genomic testing. From predesigned and validated content to custom solutions, our flexible technologies are designed to provide rapid and reliable results to responsible breeders, veterinarians and researchers. Our solutions include microarrays, targeted genotyping-by-sequencing (GBS) solutions, short tandem repeat (STR) parentage kits, and Applied Biosystems Custom TaqMan SNP Genotyping Assays.
Quick links:
AgriSeq targeted genotyping-by-sequencing (GBS)
Custom marker panels allowbreeders, veterinarians and researchers to target the most informative and important parentage, traits and defect markers for their breeds and varieties. Use in conjunction with our Applied Biosystems AgriSeq genotyping-by-sequencing (GBS) solutions to help deliver unambiguous sequence-based genotyping results.
Library Kits and Barcodes
Canine
Feline
Equine
Product
Quantity
Cat. No.
Custom Equine Genetic Traits and Parentage Panel
Varies
Contact sales representative ›
Custom marker panels allowbreeders, veterinarians and researchers to target the most informative and important parentage, traits and defect markers for their breeds and varieties. Use in conjunction with our Applied Biosystems AgriSeq genotyping-by-sequencing (GBS) solutions to help deliver unambiguous sequence-based genotyping results.
Axiom microarrays
Our array-based genotyping solutions are ideal for the identification, validation and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays or customize an array specifically for your study.
Canine
Equine
Product
# SNPs
Array Format
Cat. No.
Price
Axiom Equine Genotyping Array
670,796
96 array
550583
Contact a sales representative ›
Axiom Equine Plus Genotyping Array
670,800
96-array
550624
Contact a sales representative ›
Rabbit
Product
# SNPs
Array Format
Cat. No.
Price
Axiom Rabbit Genotyping Array
200,000
96 array
550669
Contact a sales representative ›
Short tandem repeat (STR) parentage kits
Microsatellites, also referred to as short tandem repeats (STRs), are commonly used for parentage testing, identification, and forensics due to their informativeness and Mendelian inheritance. Our parentage testing kits provide all of the necessary reagents for simultaneous amplification of multiple microsatellite loci in a single multiplex PCR reaction.
Canine
Equine
Custom TaqMan SNP Genotyping Assays
These assays provide a highly flexible technology for detection of polymorphisms within any genome. Create your own assays by submitting target sequences to our secure assay design pipeline using the Applied Biosystems Custom TaqMan Assay Design Tool. This pipeline has successfully generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. TaqMan Assays can be run on any of our real-time PCR instruments or used as an endpoint assay on other third-party systems, including the Array Tape platform.
Recommended reagents