protein-a-resin-mb-kits-230x195
We offer high performance affinity particles to meet your needs for antibody purification and immunoprecipitation (IP) assays in two versatile formats, affinity resin and magnetic beads. Our protein A plus agarose affinity resin for antibody purification is available in formats ranging from agarose beads, pre-packed spin columns, 96-well filter plates, and complete IgG purification kits. Our protein A magnetic beads can be used as affinity particles for antibody purification and IP methods using manual or robotic magnetic separators.
Which particle and format is right for you?
  Protein A Plus Agarose Protein A Magnetic Beads
Particle size 45–165 microns 1 micron
Maximum Linear Velocity 30 cm/hour N/A
Composition 6% cross-linked agarose Surface blocked magnetic bead
Binding capacity 34 mg human IgG/mL resin >400 µg rabbit IgG/mL beads
Available as beads
Prepacked Cartridges
-
Spin Columns
-
96 well spin plate
-
Complete Purification Kit
-
Immunoprecipitation
Co-immunoprecipitation
  Order now Order now
bulk-pod-showcase
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Guide to antibody purification
40000-antibodies