We offer high performance affinity particles to meet your needs for antibody purification and immunoprecipitation (IP) assays in two versatile formats, affinity resin and magnetic beads. Our protein L agarose affinity resin for antibody purification is available in formats ranging from agarose beads, pre-packed spin columns, 96-well filter plates, and complete IgG purification kits. Our protein L magnetic beads can be used as affinity particles for antibody purification and IP methods using manual or robotic magnetic separators.
Which particle and format is right for you?
  Protein L Agarose Protein L Magnetic Beads
Particle size
45–165 microns
1 micron
Maximum Linear Velocity
30 cm/hour
6% cross-linked agarose
Surface blocked magnetic bead
Binding capacity
5-10 mg human IgG/mL resin
1.1 mg human IgG/mL beads
Available as beads
Prepacked Cartridges
Spin Columns
96 well spin plate
Complete Purification Kit
Order now
Order now
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Guide to antibody purification