ProcartaPlex High Sensitivity Assays
Sensitivity in the femtogram range
Invitrogen ProcartaPlex high sensitivity assays are designed to measure small concentration differences in cell culture supernatants, plasma, and serum samples with 10-fold lower LLOQs (Lower Limit of Quantification) and sensitivities for all analytes in the femtogram range. Overcome the sensitivity detection limits of conventional multiplex immunoassays.
High sensitivity multiplex panels
ProcartaPlex high sensitivity panels
HS Panel Name Cat. No. Antigen
9-Plex Human Panel,
High Sensitivity
EPXS090-12199-901 IFNγ, IL-1β, IL-2, IL-4, IL-6, IL-10, IL-12 p70, IL-17A, TNFα
5-Plex Mouse Panel,
High Sensitivity
EPXS050-22199-901 IFNγ, IL-2, IL-4, IL-6, TNFα
High sensitivity simplex kits
ProcartaPlex high sensitivity simplex kits are magnetic bead sets used to detect individual analytes. They are designed to enable the addition of targets to multiplex panels or the combination of individual high sensitivity simplex kits to produce custom panels. A corresponding basic kit is required when combining simplex kits separate from any multiplex panel.
Human high sensitivity simplex kits
Simplex Cat. No. Analyte ULOQ*
(pg/mL)
LLOQ*
(pg/mL)
Inter Assay
%CV
Intra Assay
%CV
EPXS010-10283-901 GM-CSF 7330 1.80 <10 <10
EPXS010-10228-901 IFN gamma 260 0.31 <10 <10
EPXS010-10224-901 IL-1 beta 1330.0 0.32 <10 <10
EPXS010-10221-901 IL-2 490 0.44 <10 <10
EPXS010-10225-901 IL-4 560 0.86 <10 <10
EPXS010-10278-901 IL-5 2720 0.66 <10 <10
EPXS010-10213-901 IL-6 2850 1.03 <10 <10
EPXS010-10204-901 IL-8 890 0.22 <10 <10
EPXS010-10215-901 IL-10 750.0 0.18 <10 <10
EPXS010-10238-901 IL-12p70 3090.0 0.75 <10 <10
EPXS010-12017-901 IL-17A 1030.0 0.25 <10 <10
EPXS010-10281-901 MCP-1 650 0.16 <10 <10
EPXS010-10223-901 TNF alpha 1390 0.45 <10 <10
Mouse high sensitivity simplex kits
Simplex Cat. No. Analyte ULOQ*
(pg/mL)
LLOQ*
(pg/mL)
Inter Assay
%CV
Intra Assay
%CV
EPXS010-20606-901 IFN gamma 1250.0 0.31 <10 <10
EPXS010-20601-901 IL-2 1820.0 0.44 <10 <10
EPXS010-20613-901 IL-4 3530.0 0.86 <10 <10
EPXS010-20603-901 IL-6 4230.0 1.03 <10 <10
EPXS010-20607-901 TNF alpha 1830.0 0.45 <10 <10
*determined for first lot
Connect with us
Visit this link to answer all your questions when new to our multiplex technologies or if you need to troubleshoot unexpected results. Additionally guides and tools including videos are available for easy learning.
Email us to receive one to one consultation with a Luminex Field Application Scientist who can provide answers to determining the right products, product use and recommendations for data analysis.
Performance guarantee
If an Invitrogen immunoassay does not perform as described on our data sheet, we will replace the product at no cost to you, or if you prefer, we will provide you with a credit for future purchase.*
Learn more about performance guarantee
We're here to help
Our team of field application specialists are trained in immunoassay workflows. We can help you choose an assay or configure a panel, optimize an experiment, troubleshoot and more – face to face or on the phone at no cost.
Contact a field application specialist