Search from over 1000 ELISA and ProQuantum immunoassay kits
ProQuantum immunoassays for immuno-oncology research
ProQuantum high-sensitivity immunoassays for immuno-oncology research: ultra-low detection and ultra-low volume requirements
Cytokines are immune cell messengers that stimulate immune effector cells and stromal cells in tumors and also increase the recognition of tumor cells by cytotoxic effector cells. Investigations using animal models have revealed that cytokines display anti-tumor activity, resulting in an increased interest in the use of cytokines as a basis for cancer therapy. Many cytokines, such as GM-CSF, IL-7, IL-12, IL-15, IL-18 and IL-21, are in clinical trials for patients with advanced cancers. Pre-clinical research has focused on being able to neutralize cytokines that act to suppress the immune response, for example IL-10 and TGF-β. Adoptive cell therapy is an approach that makes modification to T cells to focus them on killing tumor cells, while controlling the cytokine response for optimal effect.
In addition, two cytokines are currently approved by the FDA for the treatment of melanoma (IL-2) and renal cell carcinoma (IFN-α). With these successes though, there continues to be a need to study the mechanisms of cytokine action in both activation and suppression of the immune system in order for them to be leveraged for the development of new anti-tumor treatments and immunotherapies against cancer.
Cytokine Primary cell source Primary target cell Biological activity
GM-CSF T cells, macrophages, endothelial cells, fibroblasts, mast cells Bone marrow, progenitor cells, DC, macrophages
NKT cells
Promotes antigen presentation, T cell homeostasis, hematopoietic cell growth factor
IL-1 Monocytes, macrophages, fibroblasts, epithelial cells, endothelial cells, astrocytes T cells, B cells, endothelial cells, hypothalamus, liver Co-stimulation, cell activation,
inflammation, fever, acute phase reactant
IL-2 T cells, NK cells T cells, NK cells, B cells, monocytes Cell growth/ activation
IL-3 T cells Bone marrow progenitor cells Cell growth and differentiation
IL-4 T cells T cells, B cells Th2 differentiation, cell growth/activation, IgE isotype switching
IL-5 T cells B cells, eosinophils Cell growth/activation
IL-6 T cells, macrophages, fibroblasts T cells, B cells, liver Co-stimulation, cell growth/ activation, acute phase reactant
IL-7 Fibroblasts, bone marrow stromal cells Immature lymphoid, progenitors T cell survival, proliferation, homeostasis, B cell development
IL-8 Macrophages, epithelial cells, platelets Neutrophils Activation, chemotaxis
IL-10 Th2 T cells Macrophages, T cells Inhibits antigen-presenting cells, inhibits cytokine production
IL-12 Macrophages, NK cells T cells Th1 differentiation
IL-15 Monocytes T cells, NK cells Cell growth/activation,NK cell development, blocks apoptosis
IL-18 Macrophages T cells, NK cells, Bcells Cell growth/activation,inflammation
IL-21 CD4+ T cells, NKT cells NK cells, T cells, B cells Cell growth/activation, control of allergic responses and viral, infections
IL-23 Antigen-presenting cells T cells, NK cells, DC Chronic inflammation, promotes Th17 cells
IFN-α Plasmacytoid DC, NK cells, T cells, B cells, macrophages, fibroblasts, endothelial cells, osteoblasts Macrophages, NK cells Anti-viral, enhances MHC expression
IFN-γ T cells, NK cells, NKT cells Monocytes, macrophages, endothelial cells, tissue cells Cell growth/activation, enhances MHC expression
TGF-β T cells, macrophages T cells Inhibits cell growth/activation
TNF- α Macrophages, T cells T cells, B cells, endothelial cells, hypothalamus, liver Co-stimulation, cell activation, inflammation, fever, acute phase reactant
References: S Lee and K Margolin, Cytokines in Cancer Immunotherapy Cancers (Basel). 2011 Dec; 3(4): 3856–3893.1.
Detect low levels of target-specific protein in immuno-oncology research
Invitrogen ProQuantum immunoassays are designed for easy-to-run, high-performance, protein detection, with no proprietary instrument to purchase. Utilizing proximity-based amplification technology, ProQuantum assays offer the analyte specificity of antibody–antigen binding with the signal detection and amplification capabilities of qPCR to achieve highly sensitivity. ProQuantum assays can also typically detect lower levels of protein with lower sample consumption than traditional methods. This can be particularly important when handling very limited volumes of precious samples from blood or tumor extracts. Limited sample volumes also present a challenge to detect very low levels of protein, but ProQuantum’s technology solves that by amplifying the signal beyond the capability of other available methods.
Whether conducting discovery or translational research to define the role and potential use of these cytokines and chemokines as targets in immunotherapies, the ability to measure precise differences at very high sensitivity levels with very small sample volumes is valuable. The next generation, ProQuantum high-sensitivity immunoassays, help immuno-oncology researchers to detect lower levels of protein with minimal consumption of precious samples than traditional methods.
See how ProQuantum immunoassays work
Learn about how immuno-oncology ELISA kits measure immune-related targets in serum, plasma and tissue culture supernatant.
Immuno-oncology ELISA kits
Watch the on-demand webinars to get the technical details of ProQuantum immunoassays from an R&D scientist.
Watch webinar
Review available ProQuantum immunoassay targets, or have us create a custom ProQuantum target for your next research project. Email us at invitrogenimmunoassays@thermofisher.com and we will connect you with a representative who can help.
Connect with us
Request the next target release
We are continuously developing new ProQuantum immunoassay kits. You can contribute to our product development roadmap. Send us an email to let us know what protein targets you would like to see next.
Send an email
Sign up for product release announcements
We are continuously releasing new ProQuantum immunoassay kits. Stay in the loop and sign up to receive our new product release announcements.
Sign up
Performance guarantee
If an Invitrogen immunoassay does not perform as described on our data sheet, we will replace the product at no cost to you, or if you prefer, we will provide you with a credit for future purchase.*
Learn more about performance guarantee
We're here to help
Our team of field application specialists are trained in immunoassay workflows. We can help you choose an assay or configure a panel, optimize an experiment, troubleshoot and more – face to face or on the phone at no cost.
Contact a field application specialist