Figure 4. Superior western blotting results obtained using ABfinity™ antibody. The STAT4 ABfinity and polyclonal antibodies were used at 2, 1, and 0.5 μg/mL. STAT4 antibodies from other vendors were used at their recommended concentrations. Primary antibody was detected using WesternBreeze Chemiluminescent Kit–anti-Rabbit.
Recombinant Rabbit Monoclonal Antibodies
Invitrogen™ rabbit recombinant antibodies are derived from rabbit monoclonal antibody-producing cell lines by isolating and cloning the specific antibody heavy and light chain DNA sequences. These recombinant cloned antibodies are not susceptible to cell-line drift or lot-to-lot variation, thus allowing for peak specificity and performance.
Our Invitrogen™ ABfinity™ recombinant rabbit antibodies are developed either as oligoclonal antibodies (clone library used for final production) or monoclonal antibodies (one clone isolated from library used for final production).
ABfinity rabbit recombinant monoclonal antibodies offer:
- Better specificity and sensitivity compared to standard antibodies
- Lot-to-lot consistency due to recombinant technology
- Animal origin-free formulations, the result of production by cell culture
How to find an ABfinity antibody for a specific target of interest
- Step 1: Search for the target of interest using the search tool below.
- Step 2: Filter results for “Rabbit” as the host (in side bar).
- Step 3: Filter result for “Monoclonal” or “Oligoclonal” (in side bar).
Need a custom recombinant rabbit monoclonal antibody? Request a quote
What is ABfinity technology?
ABfinity recombinant antibodies are highly specific and high-quality monoclonal antibodies, produced by proprietary technology, and unmatched for producing consistent results.
ABfinity antibodies are recombinant antibodies developed by immunizing animals, screening for functionality, and cloning the immunogen-specific antibody genes into high-level expression vectors. The antibodies are produced on a large scale by expressing them in mammalian cells, and purifying them with protein A. These recombinant antibodies are expressed in mammalian expression systems, but appear just like their counterparts isolated from serum or produced by hybridomas. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain.
Figure 1. Recombinant rabbit monoclonal antibody production.
ABfinity antibodies are manufactured by transfecting mammalian cells with heavy and light chain antibody cDNA. This highly reproducible process results in unparalleled lot-to-lot consistency. This consistency saves time and money because experimental conditions do not require revalidation. Figure 1 shows the consistent western blotting results achieved using independent lots of an ABfinity antibody, and its graphical representation. Figure 2 shows the lot-to-lot consistency in immunocytochemistry, and its graphical representation. Figure 3 shows the same results in flow cytometry.
![]() |
![]() |
Figure 1. Lot-to-lot consistency using western blot. HeLa cell extracts separated on reducing gels were probed with four different lots of SMAD2 ABfinity Antibody. The concentrations in lanes A, B, and C were 2 μg/mL, 1 μg/mL, and 0.5 μg/ mL, respectively. Primary antibody was detected using WesternBreeze Chemiluminescent Kit–Anti-Rabbit. Left is quantitative representation of four lots at different concentrations, showing best lot-to-lot consistency (n=4, Error bars=standard error).
![]() |
![]() |
Figure 2. Lot-to-lot consistency using immunocytochemistry. Tissue cultured HeLa cells stained with four different lots of SMAD2 ABfinity Antibody. A through D represents Lots 1 through 4 at concentration of 3 µg/mL, E through H represents Lots 1 through 4, at concentration of 0.5 μg/mL. Alexa Fluor 488 goat anti-rabbit IgG at 1:1000 was used as secondary antibody. Left is quantitative representation, of four lots at different concentrations, showing best lot-to-lot consistency (n=4, Error bars=standard error).
![]() |
Figure 3. Lot-to-lot consistency using flow cytometry. Quantitative representation of the flow cytometry, with four independent lots of SMAD2 ABfinity Antibody at various concentrations, showing high lot-to-lot consistency. HeLa cells were fixed and permeabilized using FIX & PERM Reagents. SMAD2 ABfinity Antibody incubation was followed by Alexa Fluor 488 goat anti-rabbit IgG (n=4, Error Bars= Standard Error). |
Reliable Sensitivity and Specificity
The ABfinity platform allows production of antibodies that are more sensitive and specific than those achieved with any other antibody development platform. These antibodies only react with the target of choice, eliminating detection of the wrong signal due to unspecific binding. More highly sensitive antibodies can detect very low-level targets that may be difficult to detect with other antibodies. Plus, precious samples are saved, needing less antibody for detection.
Figure 4 shows a direct comparison of an ABfinity STAT4 antibody with the best commercial STAT4 antibodies in western blotting. This comparison includes antibodies from polyclonal, traditional hybridoma monoclonal, and rabbit hybridoma monoclonal platforms. Figure 5 demonstrates the same results in immunocytochemistry. Flow cytometry (results not shown) revealed similar results, indicating the sensitivity and specificity of ABfinity antibodies.
Figure 5. Enhanced immunocytochemistry results obtained using ABfinity antibody. Immunocytochemistry results while using STAT4 ABfinity antibody at 2.5 μg/mL compared with the same antibodies as in Figure 4. STAT4 antibodies from other vendors were used at their recommended concentrations. Alexa Fluor 488 goat anti-rabbit IgG at 1:1000 was used as secondary antibody.
Antibody validation
ABfinity antibodies are validated and characterized by multiple applications. This extensive validation process allows confidence in target specificity, without any need for optimization. Examples of their application are shown in Figures 6
through 9.
|
|
|
|
||
|
|
Need antibodies in bulk quantities or custom packaging?
Inquire now. Complete Request Form
Want us to produce a custom antibody for you?
Learn more. Custom Antibody Services
For Research Use Only. Not for use in diagnostic procedures.