The His tag is a series of six to nine histidine residues generally fused to either the C- or N-terminus of a recombinant protein. The small size of the His tag, compared with other common epitope tags, makes it less likely to obstruct the target protein’s structure or function and more suitable to use under denaturing conditions. The string of histidine residues binds to several types of immobilized metal ions, including nickel, cobalt, and copper. The binding to metal ions under specific buffer conditions, allows for the simple purification and detection of His-tagged proteins.
Anti-His tag antibodies provide another dependable method for the detection and purification of tagged target proteins without a protein-specific antibody or probe. Invitrogen™ His tag antibodies are designed to reliably detect recombinant His-tagged proteins. Each antibody is validated for use in applications such as ELISA, western blot, flow cytometry, immunofluorescence, immunohistochemistry, immunocytochemistry, and immunoprecipitation. Our His tag antibodies also come with the option of being HRP, biotin, or fluorophore conjugated to meet many research needs.
Other popular His tag antibodies
SKU Cat. No. Invitrogen™ Antibody Name Size
372900 37-2900 6x-His Epitope Tag Antibody (4A12E4) 100 µg
PA19531 PA1-9531 6x-His Epitope Tag Antibody 200 µg
710286 710286 6x-His Epitope Tag Antibody (21HCLC), ABfinity™ Rabbit Oligoclonal 100 µg
MA121315D680 MA1-21315-D680 6x-His Epitope Tag Antibody, DyLight 680 conjugate (HIS.H8) 50 µL
PA1983B PA1-983B 6x-His Epitope Tag Antibody 200 µg
MA121315HRP MA1-21315-HRP 6x-His Epitope Tag Antibody, HRP conjugate (HIS.H8) 50 µL
MA121315BTIN MA1-21315-BTIN 6x-His Epitope Tag Antibody, Biotin conjugate (HIS.H8) 50 µL
MA121315 MA1-21315 6x-His Epitope Tag Antibody (HIS.H8) 100 µg
MA14806 MA1-4806 6x-His Epitope Tag Antibody (13/45/31-2) 200 µg
PA123024 PA1-23024 6x-His Epitope Tag Antibody, HRP conjugate 100 µg
Featured product data
Western blot analysis of 6x-His Epitope Tag. Analysis was performed by loading various amounts of E. coli lysate, containing a multi-epitope tagged protein, per well onto a 4–20% Tris-HCl polyacrylamide gel. Proteins were transferred to a PVDF membrane and blocked with 5% BSA/TBST for at least 1 hour. The membrane was probed with an HRP-conjugated 6x-His epitope tag monoclonal antibody (Cat. No. MA1-21315-HRP) at a dilution of 1:1,000 overnight at 4oC on a rocking platform and washed in TBS-0.1% Tween™ 20. Chemiluminescent detection was performed using Thermo Scientific™ SuperSignal™ West Pico Chemiluminescent Substrate (Cat. No. 34080).
Flow cytometry analysis of Histidine tag (6x-His) done on HEK-293 cells transiently overexpressing His-IGFBP6. Cells were fixed with 70% ethanol for 10 minutes, permeabilized with 0.25% Triton™ X-100 for 20 minutes, and blocked with 5% BSA for 1 hour at room temperature. Cells were labeled with histidine (6x-His) mouse monoclonal antibody  (Cat. No. 372900; red histogram) or with mouse isotype control (pink histogram) at a dilution of 1:400 in 2.5% BSA. After incubation at room temperature for 3 hours, the cells were labeled with Alexa Fluor™ 488 conjugated rabbit anti-mouse secondary antibody (Cat. No. A11059) at a dilution of 1:400 for 30 minutes at room temperature. The representative 10,000 cells were acquired and analyzed for each sample using an Applied Biosystems™ Attune™ NxT Cytometer. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control.
Immunofluorescence analysis of Histidine tag (6x-His) done on HEK-293 cells. Left: Untransfected control; right: anti-His (red) on His-tagged fusion proteins in HEK293 cells. The cells were labeled with 6x-His epitope tag monoclonal antibody (Cat. No. MA1-21315).  Both were counterstained with DAPI (in blue.)
Annotated product references
  • Cat. No. MA1-21315 was used in western blot to establish a process for producing HIV-1 capsid protein p24 in E.coli
    Teow SY, Mualif SA, Omar TC et al. (2013) Production and purification of polymerization-competent HIV-1 capsid protein p24 (CA) in NiCo21 (DE3) Escherichia coli. BMC Biotechnol 13:107.
  • Cat. No. MA1-21315 was used in western blot to study the ability of Escherchia coli YmdB to regulate biofilm formation independently of ribonuclease III
    Kim T, Lee J, Kim KS (2013) Escherichia coli YmdB regulates biofilm formation independently of its role as an RNase III modulator. BMC Microbiol 13:266.
  • Cat. No. MA1-21315 was used in western blot to express and purify the nucleocapsid protein of Schmallenberg virus, and raise a monclonal antibody against it
    Zhang Y, Wu S, Wang J et al. (2013) Expression and purification of the nucleocapsid protein of Schmallenberg virus, and preparation and characterization of a monoclonal antibody against this protein. Protein Expres Purif 92:1–8.
  • Cat. No. MA1-21315 was used in western blot to characterize SUMO cycle components in Hydra
    Khan U, Mehere P, Deivasigamani S et al. (2013) The Hydra small ubiquitin-like modifier. Genesis 51:619–629.