Gibco® FoamAway™ Irradiated AOF is an animal origin-free antifoaming/defoaming agent for use in cell culture systems. FoamAway™ has a sterility assurance of 10-6 and is available in ready-to-use packaging or customized for your specific application.
FoamAway™ Irradiated Product List
Speak with a customer service representative: (800) 955-6288, ext. 46966
Customize your FoamAway™ Packaging
Order Now
Bag Size and
Fill Volume
Bag Size Fill Volume (L) Fill Volume (% of bag size) Standard Increments
250mL 0.25 100% N/A
1L 0.3 - 1 30 - 100% 10% or 100mL
5L 1.5 - 5 30 - 100% 10% or 500mL
10L 4 - 10 40 - 100% 10% or 1L
20L 6 - 10 30 - 50% 10% or 2L
Option A. 250mL / 1L bag with 3 base ports
  Tube Diameter
(ID x OD)
Tube Length Tube Type Connector Type
Base Port 1 Fill port for internal use only
Base Port 2 1/4" ID 3" PVC Spike port
Base Port 3 1/4"x3/8" 3" C-flex Female luer adapter, male septum cap
Option B: 5L / 10L / 20L bag with 4 base ports
  Tube Diameter
(ID x OD)
Tube Length Tube Type Connector Type
Base Port 1 1/8" x 1/4" 40" C-flex Female luer adapter
Base Port 2 3/8" x 5/8" 40" C-flex MPC quick connect
Base Port 3 1/4" ID 3" PVC Spike port
Base Port 4 3/16" x 5/16" 40" C-flex Male luer adapter, female cap
Tube Diameter, Material and Connector Options
Tube Diameter
(ID x OD)
Tube Material Options Connector Options
.125" x .255" C-flex, Pharmed, Silicone Threaded male and female luers
1/8" x 1/4" C-flex, Pharmed, Silicone Threaded male and female luers
.151" x .215" PVC Threaded male and female luers
3/16" x 5/16" C-flex, Pharmed, Silicone Threaded male and female luers; blood spikes
1/4" x 7/16" C-flex, PVC, Silicone Threaded male and female luers; MPC quick connects; blood spikes; Tri-clover clamps
3/8" x 5/8" C-flex, Pharmed, Silicone All MPC quick connects; Kleenpak