Committed to Perfecting Cell Culture at Every Scale
Perfect your cell culture processes at every stage—from research and process development to large-scale biomanufacturing
When it comes to producing consistent quality vaccines and biologics designed to improve and save lives, nobody can do it alone. We have the hands-on expertise to help you accelerate productivity and ease the burden of regulatory compliance. With a comprehensive range of cell production and storage solutions, a global supply chain and commitment to service and support, we’re with you every step of the way.
The quality of our production cell culture platform, Thermo Scientific™ Nunc™ Cell Factory™ Systems, reflects the rigorous standards that have made us the world's leading provider of cell culture products.
Nunc High Density Cell Factory System offers
Thermo Scientific™ Nunc™ High Density Cell Factory™ System offers 30% more surface area and yield* than the standard Cell Factory system or similar multi-tray systems for adherent cell culture. Its economic benefits are immediate with impacts across many functions of an organization, as it enables end users to optimize their cell culture manufacturing footprint, reduce material usage and labor-consumption as well as to increase their cell culture yield.
*The increase in yield may vary depending on the type of cells cultured.
Download brochure
Featured Nunc Cell Factory Systems
Nunc High Density Cell Factory System
Nunc High Density Cell Factory System offers 30% more surface area and yield* all within a standard Cell Factory system footprint.  It is an efficient, economic option for increasing production capacity and cell culture yields.  Constructed of the same materials and cell culture surface as a standard Cell Factory system, it supports the continued use of current protocols and processes and minimizing the need for change.
Features:
  • Increase your Yield
    30% MORE surface area and yield* all within the standard Nunc Cell Factory system footprint
  • Enhance your Productivity 
    Increase your labor and handling efficiencies by achieving more output in a single process run
  • Improve your Process Economics
    Increase your manufacturing capacity without capital investment
  • Seize Environmental Opportunities
    Consume less and reduce your decontamination and waste disposal costs
*The increase in yield may vary depending on the type of cells cultured.
lot-to-lot consistency with Nunc Cell Factory Systems
A proven solution for large-scale production of cells, vaccines, and cellular therapeutics, these systems have the same growth kinetics as laboratory-scale cell culture products. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling, and harvesting.
Save valuable space with EasyFill Cell Factory systems
Each 10-layer, single-use system holds the equivalent of 36 T175 flasks. The linear multi-layer cell culture format is easy to use and promotes easy scalability, ideal for both research and commercial-scale applications.
A full line of Nunc Cell Factory accessories
Supports the use of aseptic methods for fluid handing relating to filling, inoculation, feeding, and harvesting of the Cell Factory system. These accessories enable the further development of applications requiring a more closed cell culture system, reducing the number of open interventions.
commercial scale process
Supports the use of aseptic methods for fluid handing relating to filling, inoculation, feeding, and harvesting of the Cell Factory system. These accessories enable the further development of applications requiring a more closed cell culture system, reducing the number of open interventions.
Related products
Thermo Scientific™ Nunc™ Automatic Cell Factory™ Manipulator System (ACFM)
Simplify and achieve constant and reliable processes. A fully programmable manipulator that automates the handling of Nunc Cell Factory Systems. Designed for use in clean room environments, all functions can be activated by the touch of a button.
Thermo Scientific™ Nunc™ Cell Factory™ Incubators
A mobile solution for incubation of 16 x Cell Factory 40 or 48 x Cell Factory 10 Systems. Available with and without CO2 control. Easily load and unload carts with the convenient floor height of the Incubators.
Thermo Scientific™ Nunc™ Cell Factory™ Shaker System
Eliminate traditional need for shaking and pounding with the Nunc Cell Factory Shaker System. The shaker system offers an automated solution for the detachment of cells from Nunc Cell Factory Systems.
Thermo Scientific™ Nunc™ Cell Factory™ Racks and Carts
Manipulate, incubate and transport. Thermo Scientific Nunc Cell Factory systems effectively using Nunc Cell Factory Racks and Carts. Each rack can hold up to four Cell Factory 40 or 12 Cell Factory 10 systems. Racks and carts are designed to integrate perfectly with all Nunc Cell Factory equipment.
Automated Cell Factory Equipment
How to Use Nunc Cell Factory Systems