Whether your cleaning needs are for high volumes or smaller quantities, Thermo Scientific™ critical container processing and cleaning services have the flexibility to meet your cleaning requirements.
We have the capability to clean plastic or glass containers up to 50L. Choose from the wide variety of Thermo Scientific containers or send us your own to process.
Containers for critical environment applications are processed and certified to each customer's specifications. To see how easily your custom application can become routine, fill in our custom cleaning specification sheet and receive feedback from our customer service specialists.
Request a quote for your custom processing needs ›
Processing and Cleaning Service Critical containers
Recommended for biopharma, vaccine and quality control applications
All cleaning services and packaging can be done in our certified class 100/10 clean room with full traceability. Thermo Scientific processes available for standard and custom containers include:
  • Low particle cleaning
  • Depyrogenation
  • Chemical cleaning for trace analysis
  • Nuclease-free cleaning and certification
  • Custom sterilization
  • Surface modification services
  • Custom packaging services
  • Class 100/10 cleaning services
Processes available for standard and custom containers
Low particle cleaning
Low particle processing is designed for use in any clean room applications, exceeding USP requirements, with validated claims lower than 1/3 the allowable particulates according to USP 788 specifications. Containers and closures are cleaned in proprietary, specially designed, HEPA-filtered washing/drying equipment in our Class 100 cleanrooms and clean-packaged in Class 10 HEPA filtered workstations.
Depyrogenation
Low endotoxin containers are designed for use in packaging when contents will be terminally sterilized or in lab environments where endotoxin content must be carefully limited or eliminated. Containers are cleaned in proprietary HEPA-filtered washing equipment with endotoxin-free water, depyrogenated at 250°C, and clean-packaged in HEPA-filtered workstations located inside our Class 100/10 clean rooms.
Chemical cleaning for trace analysis
You have your choice of a combination of several different cleaning methods developed for removal of trace inorganic, trace organic, volatile organic, or total organic carbon residues. These methods provide assurance that your packaging or analytical results are free from contaminants. Available with clean room packaging or standard packaging. Cleaning and certification services are available for many parameters, including:
  • Trace inorganics (metals)
  • Trace volatile organic compounds
  • Particle counting
  • Trace semivolatile organic compounds
  • Trace organic carbon
  • Trace pesticides/herbicides
Thermo Scientific Custom Processing Services
Save resources with custom processing, packaging and sterilization services by leaving the cleaning of your critical application containers to us. We have the capability to clean glass or plastic containers, stoppers, closures or other component parts up to 50L in size.
All services and packaging are performed in our ISO 9001 registered facility using certified Class 10 packaging areas in Class 100 clean rooms with documented traceability and GMP compliance.
Resources
Watch the Particulate Carboy Washer 10L to 50L video
  Particulate Linear Washer up to 10L video