Versatile, clean, single-use bottles
Designed specifically for the storage and transport of active pharmaceutical ingredients and bulk intermediates, Thermo Scientific™ Nalgene™ production bottles and carboys offer leak-proof storage for sensitive liquids, buffers, media, and more. They are available in PC, PETG, and HDPE, and in volumes ranging from 30 mL to 20 L. Extensive regulatory documentation assures that the products meet your demanding biotechnology requirements and reduces the time and cost to implement them in your process.
Popular production bottles and accessories
Featured production bottles and accessories categories
Our high-quality plastic Nalgene bottles are suitable for pharmaceutical and biotechnology applications that require processing and storage of bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. The materials are biologically tested and guaranteed leakproof. Select from a variety of resin materials and bottle shapes and sizes, ranging from 0.5 mL to 50 L.
Enhance the performance of your production bottle. Ensure a tight seal for your Nalgene bottles with Nalgene replacement closures and heat-shrink bands.
We are committed to partnering with our customers to help perfect their processes. Start with a simple hand sketch, a fully-detailed drawing, or anything in between. Our engineers and sales professionals will gather your input and design systems to meet your specific needs. We can support your designs or design it for you based on your specifications.
Quality, choice, and innovation for your applications
Enhance productivity through innovative general labware products. Our proven lab consumables will meet your specialized application needs, and help you maintain a clean, safe, and efficient laboratory.