Durable and built for performance
Contain and store bioproduction fluids in these durable, all-purpose vessels. Nalgene™ production tanks are economical alternatives to stainless-steel tanks. Designed for use as closed systems, they are an excellent choice for reagent storage, aseptic mixing, and dispensing. Our tanks are available in a variety of sizes and shapes, ranging from 75 L to 380 L, and are production-ready.
Popular production tanks
Designed specifically to fit Nalgene cylindrical tanks, these tank liners feature an open-bag, flat-bottom design to enhance your mixing capabilities and to maximize the utility of your carboys and tanks. 
Quality, choice, and innovation for your applications
Enhance productivity through innovative general labware products. Our proven lab consumables will meet your specialized application needs, and help you maintain a clean, safe, and efficient laboratory.