CaptureSelect™ ligands and affinity chromatography resins can be used for the highly efficient capturing step for a variety of biopharmaceuticals.
Affinity resins
  • Selectivity
    high purity in single step/feed-stock independent
  • Mild elution conditions
    retaining biological activity of target
  • Reduction of process steps
    higher yields, reduced costs
  • Efficient clearance of HCP, DNA, virus
    high selectivity in capture step
We offer products for large-scale purification of various biologics such as antibodies and antibody fragments, fusion proteins, and hormones.
We also offer complementery leakage ELISA ›.
CaptureSelect™ affinity matrices from Thermo Fisher Scientific:
Product
Target applications
Uniqueness
RoboColumn
CaptureSelect™ IgG-CH1
Human IgG antibodies and Fab fragments thereof
CH1 domain binding, no free light chain binding for fragments
CaptureSelect™ KappaXL
Human IgG antibodies and Fab fragments thereof containing a kappa light chain
Mild elution for fragments and antibodies
KappaXL
CaptureSelect™ FcXL
Human IgG antibodies and Fc-fusion proteins
CH3 domain binding, human-specific, recognizing all four subclasses, mild elution conditions
FcXL
CaptureSelect™ CH1-XL Affinity Matrix
Human IgG antibodies and Fab fragments
Ch1 domain binding , can purify IgGs and antibody fragments in the presence of fetal calf serum
POROS
MabCapture™ A Resin
Monoclonal antibody capture applications
Obtain highest dynamic binding capacity, at flow rates over 700 cm/hr
MabCapture^TM A
POROS™
MabCapture™ A Select Resin
Monoclonal antibody capture applications Major increase in process productivity and reduction in downstream bottleneck MabCapture^TM A Select
View additional CaptureSelect™ products ›
Contact a member of our business development team to discuss possibilities for further development of these products.
CaptureSelect™ information
CaptureSelect™ Leakage ELISAs
Products incorporating CaptureSelect™ technology have already been developed and are used for clinical and commercial manufacturing of a variety of biologicals. To support these products Thermo Fisher Scientific has developed product-specific Leakage ELISAs that can be used for routine quality analysis. Below is a list of Leakage ELISAs and their corresponding affinity resins.
Product Complementing resin
CaptureSelect™ IgG-CH1 Ligand Leakage ELISA
CaptureSelect IgG-CH1
CaptureSelect™ KappaXL Ligand Leakage ELISA
CaptureSelect KappaXL
CaptureSelect™ FcXL Ligand Leakage
CaptureSelect FcXL
These chromatography resins, based on the CaptureSelect™ technology, have all been developed in collaboration with GE Healthcare and are currently in use for clinical and commercial manufacturing of biological compounds.
Resources
Application notes
Articles
  • Application of a novel affinity adsorbent for the capture and purification of recombinant factor VIII compounds. McCue JT, Selvitelli K, Walker J. (2009) J Chromatogr A. 1216(45):7824-30.
  • The first recombinant human coagulation factor VIII of human origin. Casademunt E, Martinelle K, Jernberg M, et al. (2012). Eur J Haematol. 89(2):165-76.
For more information on CaptureSelect