The AGT™ process allows production of complete, complex, chemically-defined, serum-free, and protein-free media in a dry format that is easy to use and easy to scale up. The following Gibco™ catalog cell culture media are available in AGT format off-the-shelf, and your own custom formulation can be manufactured in AGT format on a make-to-order basis.
Gibco™ FunctionMAX™ TiterEnhancer
A revolutionary additive for Chinese hamster ovary (CHO) fed-batch processes.
  • Up to double titer
  • Highly concentrated, pH-neutral additive
  • Minimal optimization required
Gibco™ EfficientFeed™ + Supplements
Fast, flexible performance in fed-batch cell culture.
  • Superior performance
  • Reconstitution flexibility up to 3X concentration
  • pH-neutral addition
Gibco™ Dynamis™ AGT™ Medium
The power to achieve higher titers, start faster, and streamline scale up.
  • Superior industry-leading performance
  • pH-neutral addition
  • Animal origin-free
  • Chemically defined
  • Protein free
Gibco™ CD OptiCHO™ AGT™ Medium
Optimized for the growth of CHO cells and expression of recombinant proteins in suspension culture.
  • Animal origin free
  • Chemically defined
  • Protein free
Gibco™ CD CHO AGT™ Medium
Optimized for the growth of CHO cells and expression of recombinant proteins in suspension culture.
  • Animal origin-free
  • Chemically defined
  • Protein free
Gibco™ CD Hybridoma AGT™ Medium
Optimized for the growth of a variety of hybridoma and myeloma and the production of monoclonal antibodies in stationary or agitated suspension systems.
  • Animal origin-free
  • Chemically defined
  • Protein free
Technical information
Gibco™ VP-SFM AGT™ Medium
Optimized for the growth of VERO cells for virus production; also suitable for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells and for recombinant protein production.
  • Animal origin-free
  • Serum free
  • Ultra-low protein (5µg/mL recombinant protein)
Technical information
Gibco™ CD 293 AGT™ Medium
Optimized for the growth of suspension cultures of 293 cells.
  • Animal origin-free
  • Chemically defined
  • Protein free
Technical information
Custom AGT™ Products
We can produce cGMP modifications of Gibco™ catalog media or your own complete custom formulations in AGT format, including chemically-defined, serum-free, and protein-free media. Some of our most popular formulations, such as Gibco™ OptiPRO™ SFM, are also available in AGT format by request.
Learn more about customized cGMP media ›
Non-cGMP Feasibility Batches
We can produce non-cGMP feasibility batches in AGT format using our small-scale fluid bed granulator in batch sizes up to 8 kg.  Formulations can include modifications of Gibco catalog media or your own complete custom formulations, including chemically-defined, serum-free and protein-free media.
Learn more about non-cGMP media from Gibco™ MediaExpress™ ›