Our global manufacturing capabilities ensure your access to cGMP facilities for the production and testing of recombinant protein therapeutics and cell therapies, as well as bulk media manufacturing by cell culture industry leader Gibco®.
Request Information
CMO Partnerships
If you need an integrated development/manufacturing solution, Invitrogen's Gibco® Bioprocess Services brings additional value to your programs through established partnerships with pre-qualified contract manufacturers.  This partnership approach allows Invitrogen to focus on solving your process development challenges through our technology expertise, while also bringing value through our CMO partners' strengths in cGMP manufacturing execution.  This approach gives you more choice in project timing and process options by making several different facilities available for cGMP manufacturing.  We work with each client to ensure that the right contractor is selected based on the specific requirements of your project – technical capabilities, scheduling capacity, pricing flexibility, geographical location or any combination thereof.
Custom Cell Culture Media & Services
Choose Gibco® Cell Culture Custom Media Services for large-scale cGMP media production.  We can produce liquid media in volumes up to 10,000 L with packaging options up to 1,000 L.  Dry powder and AGT™ formats are available for batches up to 7,500 kg (dependent on the bulk density of the powder).
Learn more about Custom Cell Culture Media & Services
Packaging Solutions
You have the option of receiving your Gibco® custom media in the packaging that best suits your needs.  We have many different options for liquid and powder media in a variety of package sizes, including small, intermediate and large scale.
Learn more about Custom Packaging Options
AGT™ Dry Format Media
AGT™ (Advanced Granulation Technology™) is a dry media format that allows for complete serum-free formulations and ensures high consistency and productivity across all stages of production from development to commercial manufacturing.
Learn more about AGT™ media