Critical Environment Containers
The only thing in your container is what
you add
Working within a critical environment necessitates high-quality materials, as contamination can quickly spell disaster for what are typically exceedingly sensitive applications.
How do you select the best containers for your application? Container cleanliness is critical. Understanding different cleanliness and validation options, as well as their impact, can help you to identify quality products that will not influence the integrity of your product.
Featured Critical Environment Container Categories
Process definitions table
With the capabilities to generate a broad combination of cleanliness parameters and certifications, coupled with custom packaging options, we can meet your precise specifications and requirements.   Download table ›
Process Definition
Irradiation
The application of a dose of radiation in an attempt to destroy all viable forms of life, including bacterial spores. The typical radiation dose required to reach the desired sterility assurance level (SAL) of 10–6 is 25–40 kGy. Simply being “irradiated” is not a guarantee of sterility (see “Sterile” below).
Autoclaving
The application of steam, heat, and pressure in an attempt to destroy all viable forms of life, including bacterial spores. The amount of time, heat, and pressure can vary depending upon several factors. Simply being “autoclaved” is not a guarantee of sterility (see “Sterile” below).
Sterile
Sterilization involves the application of steam, heat, and pressure (autoclaving), irradiation (e.g., gamma radiation, electron beam), or chemicals (e.g., EtOH, H2O2) to destroy all viable forms of life, including bacterial spores, to an acceptable sterility assurance level (SAL) of 10–6 that has been fully validated. Validation generally includes a statistically significant sampling plan, multiple lots, and the use of validated equipment and processes.
Particulate
cleaning
Water filtered to submicron levels (USP water or WFI, for example) is used for the particulate-cleaning process. Water is heated and used to “wash” items in multiple cycles. Materials are then dried and packaged, all in clean-room environments to keep particulates low. Simply being “particulate washed” is not a guarantee of sterility (see “USP <788>” below).
USP <788>
Compliance to USP <788> requires that either a product is produced in a clean environment or a product is particulate washed, such that you can achieve a validation of this low-particulate limit. Validation generally includes a statistically significant sampling plan, multiple lots, and the use of validated equipment and processes.
Related products and services
Ordering information
Low particulate glassware and plasticware
Depyrogenated glassware
Total organic carbon glassware & water
Silanized Glassware
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: