Single-Use Benchtop Bioreactor
Partnering high performance and simplicity to yield exceptional cell growth
The Thermo Scientific HyPerforma Single-Use Benchtop Bioreactor provides a plug-and-play solution for process development and research applications. When you require a straightforward, single-use vessel, the HyPerforma Single-Use Benchtop Bioreactor is suitable for applications that require either product sensitivity, or more robust, stirred-tank mixing capabilities.
The single-use vessel eliminates the need to autoclave, is quick to set-up-ready-to-use and provides scalability of a stirred-tank design. The bioreactor is equipped with a Thermo Scientific TruFluor pH+dO2 single-use sensor to monitor product parameters, as well as an embedded optical reader—easily attached and connected to the controller for instant recognition by TruBio software.
Request information
Key Features of Single-Use Benchtop Bioreactors
  • Shipped gamma-irradiated and ready for use.
  • Latex-free, phthalate-free, BPA-free, animal component–free materials
  • Heating blanket includes bimetallic temperature- limiting switch embedded in the blanket to help protect against overheating or fires
  • Optional customization: configure head plate, impellers, and sparger for intended use*
  • Drilled pipe (A) at bottom of vessel positioned to ensure a high turndown ratio
  • pH and dissolved oxygen (DO) sensor (B)
  • Attach the reader (C), and vessel is instantly recognized by TruBio software
  • Additional components, such as optical or electrochemical probes (D), can be installed into the configurable head plate with the use of a clean bench
* Customization feasibility may need additional assessment.
Single-Use Benchtop Bioreactor Key Features
Related products
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.