To download one of our application notes, please register through the link in the respective note below.
Scale-up of microbial fermentation using recombinant E. coli to produce an 80 kDa protein in Thermo Scientific™ HyPerforma™ 30 L and 300 L Single-Use Fermentors
Introduction:
Fermentation production facilities can utilize the Thermo Scientific™ HyPerforma™ Single-Use Fermentor (S.U.F.) instead of traditional stainless steel cleaning-in-place/sterilization-in-place (CIP/SIP) fermentor vessels without modifying their existing procedures. As expression systems are moved to production volumes, they can easily be scaled from a benchtop fermentor (or flask) to the S.U.F. Unlike hybrid solutions—cell culture bioreactors converted to fermentation systems—the S.U.F. has been specifically designed to meet the rigorous demands of microbial fermentation and delivers yields that are comparable to traditional CIP/SIP systems.
Scale-up and 2,000 L production scale of CHO cells using the Thermo Scientific™ HyPerforma™ Single-Use Bioreactor
Introduction:
The stirred-tank reactor is one of the most commonly utilized reactor types in biotechnology. The Thermo Scientific™ HyPerforma™ Single-Use Bioreactor (S.U.B.) is designed as a stirred-tank reactor for animal cell culture and has the unique advantage of utilizing completely single-use product contact surfaces. Available in 25 L to 2,000 L working volumes, the S.U.B. is one of the industry’s most widely used bioreactors today.
Thermo Scientific™ Harvestainer™ BioProcess Container: microcarrier bead separation and cell harvest
Introduction:
Techniques used to separate anchorage-dependent mammalian cells from microcarrier beads include: sedimentation using conical or incline settlers, centrifugation, acoustic resonance, and spin filtration and microfiltration. These techniques often use sophisticated equipment that require a significant capital expenditure as well as routine maintenance and between-use cleaning and sterilization. The 200 L Thermo Scientific™ Harvestainer™ BioProcess Container offers a robust and economical alternative to other systems.
Thermo Scientific™ HyPerforma™ Single-Use Mixer performance report
Introduction:
Single-use systems are widely adopted in upstream and downstream bioprocessing, from media storage containers and filling manifolds to mixing vessels and bioreactors. We offer a wide range of single-use mixing solutions—from docking stations that can be used for mixing as well as product storage, to stand-alone mixing systems for dry powder and liquid, to liquid mixing of 25 L to 2,000 L working volumes.
Contact us below for any reason, such as:
  • Have a sales rep contact me
  • Have a technical field application specialist contact me
  • I would like a product quote
  • I would like a product sample
  • I would like a product demo
  • I have a question